Skip to Content
Merck
All Photos(3)

Documents

N6639

Sigma-Aldrich

Nitrotetrazolium Blue chloride

powder, electrophoresis grade

Synonym(s):

2,2′-bis(4-Nitrophenyl)-5,5′-diphenyl-3,3′-(3,3′-dimethoxy-4,4′-diphenylene)ditetrazolium chloride, 3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis[2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride], p-Nitro-Blue tetrazolium chloride, p-Nitrotetrazolium blue, NBT, Nitro BT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C40H30N10O6 · 2Cl
CAS Number:
Molecular Weight:
817.64
Beilstein:
4115923
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.52

grade

electrophoresis grade

Quality Level

Assay

>90%

form

powder

technique(s)

electrophoresis: suitable

solubility

H2O: 10 mg/mL (A stock solution at 10mg/ml in water is stable for 1-2 weeks in the dark at 0-4°C.)
2-methoxyethanol: 20 mg/mL
ethanol: 5 mg/mL

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].COc1cc(ccc1-[n+]2nc(nn2-c3ccc(cc3)[N+]([O-])=O)-c4ccccc4)-c5ccc(c(OC)c5)-[n+]6nc(nn6-c7ccc(cc7)[N+]([O-])=O)-c8ccccc8

InChI

1S/C40H30N10O6.2ClH/c1-55-37-25-29(13-23-35(37)47-43-39(27-9-5-3-6-10-27)41-45(47)31-15-19-33(20-16-31)49(51)52)30-14-24-36(38(26-30)56-2)48-44-40(28-11-7-4-8-12-28)42-46(48)32-17-21-34(22-18-32)50(53)54;;/h3-26H,1-2H3;2*1H/q+2;;/p-2

InChI key

FSVCQIDHPKZJSO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nitrotetrazolium Blue chloride (NBT) is a dye that is used for detection of alkaline phosphatase in combination with 5-bromo-4-chloro-3-indoxyl phosphate (BCIP).

Application

Nitrotetrazolium Blue chloride has been used:
  • to determine the superoxide anion in skeletal muscle cells
  • as a component in Tris-HCl buffered saline for nicotinamide adenine dinucleotide phosphate (NADPH)-diaphorase histochemistry of brain sections
  • as a chromogenic substrate to study the histochemical detection of hydrogen peroxide (H2O2) and superoxide (O2- ) anion in Brassica juncea seedlings
  • as a component in Triton X-100 solution to perform staining of coronal brain sections

Biochem/physiol Actions

Nitrotetrazolium Blue chloride (NBT) has also been used as a colorimetric indicator of bacterial infection in blood samples. NBT has also been used as a redox indicator for other enzymatic reactions. NBT produces a dark blue insoluble formazan compound while reacting with O2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Low Intensity Exercise Training Improves Skeletal Muscle Regeneration Potential
Pietrangelo T, et al.
Frontiers in Physiology, 6 (2015)
Ontogenesis of NADPH-diaphorase positive neurons in guinea pig neocortex
Liu C, et al.
Frontiers in Neuroanatomy, 9 (2015)
Histochemical detection of superoxide and H2O2 accumulation in Brassica juncea seedlings
Kumar D, et al.
Bio-protocol, 4(8), e1108-e1108 (2014)
The systemDrive: a Multisite, Multiregion Microdrive with Independent Drive Axis Angling for Chronic Multimodal Systems Neuroscience Recordings in Freely Behaving Animals
Billard MW, et al.
eNeuro, 5(6) (2018)
Chihiro Ebihara et al.
Human molecular genetics, 24(15), 4238-4249 (2015-05-03)
Seipin, encoded by BSCL2 gene, is a protein whose physiological functions remain unclear. Mutations of BSCL2 cause the most-severe form of congenital generalized lipodystrophy (CGL). BSCL2 mRNA is highly expressed in the brain and testis in addition to the adipose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service