Skip to Content
Merck
All Photos(1)

Documents

N1892

Supelco

Nadolol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO4
CAS Number:
Molecular Weight:
309.40
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CC(C)(C)NCC(O)COc1cccc2C[C@@H](O)[C@@H](O)Cc12

InChI

1S/C17H27NO4/c1-17(2,3)18-9-12(19)10-22-16-6-4-5-11-7-14(20)15(21)8-13(11)16/h4-6,12,14-15,18-21H,7-10H2,1-3H3/t12?,14-,15+/m1/s1

InChI key

VWPOSFSPZNDTMJ-UCWKZMIHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

β-Adrenergic blocker

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Besana et al.
Journal of cardiovascular pharmacology, 59(3), 249-253 (2011-10-28)
Beta-adrenergic receptor antagonists (β-blockers) are the therapy of choice for the long QT syndrome but their efficacy is not homogeneous: propranolol and nadolol are the most effective, whereas metoprolol is associated with more treatment failures. Propranolol has a blocking effect
Ling Wang et al.
The Science of the total environment, 426, 289-295 (2012-04-17)
Beta-blockers, prescribed for the treatment of high blood pressure and for long-term use after a heart attack, have been detected in surface and ground waters. This study examines the photochemical fate of three beta-blockers, atenolol, metoprolol, and nadolol. Hydrolysis accounted
Atrial fibrillation in a LQTS patient with an ICD programmed for managed ventricular pacing: what is the cause?
Robert M Gow et al.
Pacing and clinical electrophysiology : PACE, 35(3), 357-359 (2011-05-25)
Sarah Maud Fischer et al.
International journal of pharmaceutics, 422(1-2), 52-58 (2011-10-27)
The compatibility of fasted state simulated intestinal fluid (FaSSIF) in drug permeation studies employing the phospholipid vesicle-based permeation assay (PVPA) model was confirmed by a set of different integrity indicators. Neither calcein permeability nor electrical resistance were found significantly changed
Sarah Maud Fischer et al.
The Journal of pharmacy and pharmacology, 63(8), 1022-1030 (2011-07-02)
The aim of this study was to determine the influence of nonionic surfactants on drug permeability using the phospholipid vesicle-based permeation assay (PVPA), which excludes other than trans-membrane diffusion pathways. Barrier integrity was monitored both by electrical resistance and permeability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service