Skip to Content
Merck
All Photos(1)

Key Documents

N0020000

Nabumetone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-(6-Methoxy-2-naphthyl)-2-butanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O2
CAS Number:
Molecular Weight:
228.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

nabumetone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc2cc(CCC(C)=O)ccc2c1

InChI

1S/C15H16O2/c1-11(16)3-4-12-5-6-14-10-15(17-2)8-7-13(14)9-12/h5-10H,3-4H2,1-2H3

InChI key

BLXXJMDCKKHMKV-UHFFFAOYSA-N

Gene Information

human ... PTGS2(5743)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nabumetone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Nonselective non-steroidal anti-inflammatory drug (NSAID)

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Crothers et al.
International journal of pharmaceutics, 358(1-2), 303-306 (2008-04-18)
The solubilisation of two poorly soluble drugs, furosemide and nabumetone, in micellar solutions of diblock copolymers of ethylene oxide and styrene oxide has been studied at 25 and 37 degrees C and solubilisation capacities compared with published values for griseofulvin
Kaori Matsumoto et al.
Biological & pharmaceutical bulletin, 34(5), 734-739 (2011-05-03)
The cytochrome P450 (CYP) isoforms that catalyze the oxidation metabolism of 6-methoxy-2-napthylacetic acid (6-MNA), an active metabolite of nabumetone, were studied in rats and humans. Using an extractive reversed-phase HPLC assay with fluorescence detection, monophasic Michaelis-Menten kinetics was obtained for
Solubility of triamcinolone acetonide and other anti-inflammatory drugs in silicone oil: implications for Therapeutic Efficacy.
José Carlos Pastor et al.
Retina (Philadelphia, Pa.), 28(9), 1247-1250 (2008-07-17)
R Andrew Moore et al.
The Cochrane database of systematic reviews, (4)(4), CD007548-CD007548 (2009-10-13)
Nabumetone is a non-steroidal anti-inflammatory drug (NSAID) used mainly in treating pain associated with arthritis. The usual oral dose for osteoarthritis is 1000 mg daily, and higher doses are not advised in older patients. There are no known systematic reviews
Jinhyun Kim et al.
Clinical therapeutics, 31(12), 2860-2872 (2010-01-30)
S-adenosylmethionine (SAMe) has antiinflammatory and analgesic effects and has been reported to ameliorate the pain and dysfunction of osteoarthritis (OA). The metabolism of SAMe can be affected by geographic or ethnic factors. However, its efficacy and tolerability versus NSAIDs have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service