Skip to Content
Merck
All Photos(3)

Key Documents

M9546

Sigma-Aldrich

D-Mannitol

BioXtra, ≥98% (HPLC)

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

product line

BioXtra

Quality Level

Assay

≥98% (HPLC)

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.01% Insoluble matter

ign. residue

≤0.01%

color

white

useful pH range

5.0-6.5 (25 °C, 182 g/L)

mp

167-170 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-Mannitol is the alcohol form of mannose. Its molar mass is 182 g/mol. Mannitol is present widely in plants, fungi and algae. It is extracted mainly from seaweeds. It is used in food for diabetic patients as it is absorbed slowly. It is commonly used as a stabilizer, humectant, nutritive sweetener and bulking agent in food.[1]

Application

D-Mannitol has been used:
  • as a hydroxyl radical inhibitor, to detect the hydroxyl radical generation during the reactive oxygen species (ROS) modification of chromatin[2]
  • in [3H]-mannitol permeability assay and also as a component in buffer B for measuring the transmonolayer dilution potentials[3]
  • as a component in extracellular recording solution for whole cell patch clamp recordings[4]

Biochem/physiol Actions

D-Mannitol is used as an osmotic agent for the treatment of brain swellings.[5]
A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sweeteners: Nutritional Aspects, Applications, and Production Technology, 59-59 (2012)
Methods to examine tight junction physiology in cancer stem cells: TEER, paracellular permeability, and dilution potential measurements
Buchert M, et al.
Stem Cell Reviews and Reports, 8(3), 1030-1034 (2012)
Neurology of the Newborn, 459-459 (2008)
Allosteric interactions between NMDA receptor subunits shape the developmental shift in channel properties
Sun W, et al.
Neuron, 94(1), 58-64 (2017)
Antibodies against 4-hydroxy-2-nonenal modified epitopes recognized chromatin and its oxidized forms: role of chromatin, oxidized forms of chromatin and 4-hydroxy-2-nonenal modified epitopes in the etiopathogenesis of SLE
Al-Shobaili HA, et al.
Disease Markers, 33(1), 19-34 (2012)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service