Skip to Content
Merck
All Photos(3)

Documents

M2780

Sigma-Aldrich

(1R,2S,5R)-(−)-Menthol

ReagentPlus®, 99%

Synonym(s):

(−)-Menthol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Beilstein:
1902293
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

biological source

synthetic (organic)

Quality Level

vapor pressure

0.8 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

optical activity

[α]20/D −50°, c = 10 in 95% ethanol

optical purity

ee: 99% (GLC)

bp

212 °C (lit.)

mp

41-45 °C (lit.)

solubility

water: 20 mg/mL, clear, colorless

density

0.89 g/mL at 25 °C (lit.)

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1

InChI key

NOOLISFMXDJSKH-KXUCPTDWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(1R,2S,5R)-(-)-Menthol is a monoterpene.

Application

(1R,2S,5R)-(-)-Menthol may be used to synthesize:
  • chiral enantiopure 2-(1-hydroxyalkyl)pyridines, which can react with to form C3-symmetric tripodal ligands
  • menthylphosphorodichioridite, a chiral derivatizing agent for the determination of enantiomeric purity of chiral diols or diamines by 31P NMR spectroscopy
  • (2R,4S)-2,3,4,5-tetrahydro-2-(-)-menthyloxy-2-methyl-4-phenylpyrano-[3,2-c]-benzopyran-5-one, an intermediate to prepare a warfarin analog
It may also be used as a chiral auxiliary, to induce chirality while synthesizing (-)-horsfiline.
Used to prepare reagents for chiral vinylogous Darzens and Reformatsky reactions. Important chiral auxiliary employed in the resolution of acids and for stereocontrolled synthesis.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 52, 4397-4397 (1987)
A new 31P NMR method for the enantiomeric excess determination of diols and secondary diamines with C 2 symmetry.
Brunel JM and Faure B.
Tetrahedron Asymmetry, 6(9), 2353-2356 (1995)
Fieser, M.
Reagents for Organic Synthesis, 16, 203-203 (1992)
An asymmetric approach to coumarin anticoagulants via hetero-Diels-Alder cycloaddition.
Cravotto G, et al.
Tetrahedron Asymmetry, 12(5), 707-709 (2001)
Marei GI, et al.
Pesticide Biochemistry and Physiology, 103(1), 56-61 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service