Skip to Content
Merck
All Photos(1)

Key Documents

M1750000

Methylprednisolone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6α-Methylprednisolone, 11β,17α,21-Trihydroxy-6α-methyl-1,4-pregnadiene-3,20-dione, 6α-Methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, Medrol, Medrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30O5
CAS Number:
Molecular Weight:
374.47
Beilstein:
2340300
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methylprednisolone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

wet ice

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C22H30O5/c1-12-8-14-15-5-7-22(27,18(26)11-23)21(15,3)10-17(25)19(14)20(2)6-4-13(24)9-16(12)20/h4,6,9,12,14-15,17,19,23,25,27H,5,7-8,10-11H2,1-3H3/t12-,14-,15-,17-,19+,20-,21-,22-/m0/s1

InChI key

VHRSUDSXCMQTMA-PJHHCJLFSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methylprednisolone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Adrenal gland,Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juho Keski-Nisula et al.
The Annals of thoracic surgery, 95(6), 2126-2132 (2013-04-23)
Corticosteroids are widely used in pediatric open-heart surgery to reduce systemic inflammatory response and to mediate possible cardioprotective effects. However, the optimal dosing of corticosteroids is unknown and their administration varies considerably between different institutions. Forty neonates undergoing open-heart surgery
Christiaan Fokke et al.
Brain : a journal of neurology, 137(Pt 1), 33-43 (2013-10-29)
Guillain-Barré syndrome is an acute polyradiculoneuropathy with a variable clinical presentation. Accurate diagnostic criteria are essential for patient care and research, including clinical trials and vaccine safety studies. Several diagnostic criteria for Guillain-Barré syndrome have been proposed, including the recent
MMWR. Morbidity and mortality weekly report, 62(19), 377-381 (2013-05-17)
As of May 6, 2013, Michigan had reported 167 (52%) of the 320 paraspinal or spinal infections without meningitis associated with the 2012-2013 fungal meningitis outbreak nationally. Although the index patient had a laboratory-confirmed Aspergillus fumigatus infection, the fungus most
Emilio Portaccio et al.
Journal of neurology, neurosurgery, and psychiatry, 85(8), 845-850 (2014-01-10)
To assess relapses, disability progression and the role of disease modifying drugs (DMDs) in the year after delivery in women with multiple sclerosis (MS). We prospectively followed-up pregnancies occurring between 2002 and 2008 in women with MS, recruited from 21
Ornulf Paulsen et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 32(29), 3221-3228 (2014-07-09)
Corticosteroids are frequently used in cancer pain management despite limited evidence. This study compares the analgesic efficacy of corticosteroid therapy with placebo. Adult patients with cancer receiving opioids with average pain intensity ≥ 4 (numeric rating scale [NRS], 0 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service