Skip to Content
Merck
All Photos(1)

Key Documents

I0320000

Insulin (porcine)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Insulin from porcine pancreas, Insulin from hog pancreas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C256H381N65O76S6
CAS Number:
Molecular Weight:
5777.54
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

insulin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

InChI key

NOESYZHRGYRDHS-ZYCCASTOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Insulin (porcine) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.
Two-chain polypeptide hormone produced by the β-cells of pancreatic islets. Its molecular weight is ~5800 Da. The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond. Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

InChI: 1S/C256H381N65O76S6/c1-29-131(23)205(313-193(337)104-258)251(391)317-204(130(21)22)247(387)288-159(75-82-200(347)348)217(357)282-156(71-78-189(262)333)221(361)308-183-116-401-402-117-184-242(382)305-178(111-323)239(379)294-162(88-123(7)8)224(364)295-168(95-140-53-61-146(327)62-54-140)227(367)283-154(69-76-187(260)331)218(358)290-161(87-122(5)6)222(362)285-158(74-81-199(345)346)220(360)302-174(101-190(263)334)234(374)298-170(97-142-57-65-148(329)66-58-142)230(370)309-182(241(381)304-176(255(396)397)103-192(265)336)115-400-399-114-181(213(353)272-107-194(338)278-153(72-79-197(341)342)216(356)281-151(51-42-84-270-256(266)267)211(351)271-108-195(339)279-166(93-138-46-36-32-37-47-138)226(366)297-167(94-139-48-38-33-39-49-139)229(369)299-171(98-143-59-67-149(330)68-60-143)237(377)320-208(136(28)326)253(393)321-85-43-52-186(321)245(385)286-152(50-40-41-83-257)214(354)277-134(26)254(394)395)311-249(389)203(129(19)20)316-235(375)164(90-125(11)12)292-228(368)169(96-141-55-63-147(328)64-56-141)296-223(363)160(86-121(3)4)289-209(349)133(25)276-215(355)157(73-80-198(343)344)287-246(386)202(128(17)18)315-236(376)165(91-126(13)14)293-232(372)173(100-145-106-269-120-275-145)301-238(378)177(110-322)280-196(340)109-273-212(352)180(113-398-403-118-185(310-243(183)383)244(384)319-207(135(27)325)252(392)306-179(112-324)240(380)318-206(132(24)30-2)250(390)312-184)307-225(365)163(89-124(9)10)291-231(371)172(99-144-105-268-119-274-144)300-219(359)155(70-77-188(261)332)284-233(373)175(102-191(264)335)303-248(388)201(127(15)16)314-210(350)150(259)92-137-44-34-31-35-45-137/h31-39,44-49,53-68,105-106,119-136,150-186,201-208,322-330H,29-30,40-43,50-52,69-104,107-118,257-259H2,1-28H3,(H2,260,331)(H2,261,332)(H2,262,333)(H2,263,334)(H2,264,335)(H2,265,336)(H,268,274)(H,269,275)(H,271,351)(H,272,353)(H,273,352)(H,276,355)(H,277,354)(H,278,338)(H,279,339)(H,280,340)(H,281,356)(H,282,357)(H,283,367)(H,284,373)(H,285,362)(H,286,385)(H,287,386)(H,288,387)(H,289,349)(H,290,358)(H,291,371)(H,292,368)(H,293,372)(H,294,379)(H,295,364)(H,296,363)(H,297,366)(H,298,374)(H,299,369)(H,300,359)(H,301,378)(H,302,360)(H,303,388)(H,304,381)(H,305,382)(H,306,392)(H,307,365)(H,308,361)(H,309,370)(H,310,383)(H,311,389)(H,312,390)(H,313,337)(H,314,350)(H,315,376)(H,316,375)(H,317,391)(H,318,380)(H,319,384)(H,320,377)(H,341,342)(H,343,344)(H,345,346)(H,347,348)(H,394,395)(H,396,397)(H4,266,267,270)/t131-,132-,133-,134-,135+,136+,150-,151-,152-,153-,154-,155-,156-,157-,158-,159-,160-,161-,162-,163-,164-,165-,166-,167-,168-,169-,170-,171-,172-,173-,174-,175-,176-,177-,178-,179-,180-,181-,182-,183-,184-,185-,186-,201-,202-,203-,204-,205-,206-,207-,208-/m0/s1
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Socha et al.
Drug delivery, 16(8), 430-436 (2009-10-21)
Nanoparticles loaded with two different commercial insulins (Actrapid, Novorapid and based on different blends of a biodegradable polyester (poly-epsilon-caprolactone) and a polycationic non-biodegradable acrylic polymer (Eudragit RS) were characterized in vitro. The zeta potential was positive whenever Eudragit RS was
J Hans DeVries
Current diabetes reviews, 1(1), 23-26 (2008-01-29)
Insulin pump therapy enjoys a steadily growing number of users and is associated with an approximately 0.5% lower A1c as compared to flexible insulin injection therapy in type 1 diabetes patients. An important question is whether superiority of insulin pump
Kumiko Iwata et al.
Current aging science, 3(1), 26-33 (2010-03-20)
In cartilage regenerative medicine, which is highly expected in the face of our aging society, insulin is the potent factor for culture media. To secure the safety of culture media, we attempted to use medical insulin formulations, and compared their
Arnaud Agin et al.
Clinical chemistry and laboratory medicine, 44(11), 1379-1382 (2006-11-08)
In clinical studies involving rapid-acting analogues (RAAs), insulin immunoreactivity is frequently measured, including endogenous, regular insulin (RI) and RAA immunoreactivities. Such a procedure implies equivalent cross-reactivities of all insulins present in serum. Commercially available human insulin immunoassays have been widely
Morten Donsmark et al.
Diabetes technology & therapeutics, 11(9), 581-585 (2009-09-22)
Easy-to-use insulin devices are an important tool in the treatment of diabetes. In this study, injection force requirements and dosage scale displays were evaluated for NovoPen 3 and NovoPen 4 (both from Novo Nordisk A/S, Copenhagen, Denmark). To simulate 5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service