Skip to Content
Merck
All Photos(1)

Key Documents

G0400006

Glycerol monolinoleate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H38O4
CAS Number:
Molecular Weight:
354.52
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

glyceryl monolinoleate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O(CC(O)CO)C(=O)CCCCCCC\C=C/C\C=C/CCCCC

InChI

1S/C21H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h6-7,9-10,20,22-23H,2-5,8,11-19H2,1H3/b7-6-,10-9-

InChI key

WECGLUPZRHILCT-HZJYTTRNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Glycerol monolinoleate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adryana Clementino et al.
International journal of nanomedicine, 11, 6575-6590 (2016-12-21)
Along with their cholesterol-lowering effect, statins have shown a wide range of pleiotropic effects potentially beneficial to neurodegenerative diseases. However, such effects are extremely elusive via the conventional oral administration. The purpose of the present study was to prepare and
Samuel Guillot et al.
Langmuir : the ACS journal of surfaces and colloids, 26(9), 6222-6229 (2010-02-11)
The internal phase of monolinolein-based dispersions loaded with tetradecane or (R)-(+)-limonene was investigated as a function of the stabilizer content by small-angle X-ray scattering. Phase transitions at the colloidal scale were found in some of nanostructured aqueous dispersions by increasing
Renata Negrini et al.
Langmuir : the ACS journal of surfaces and colloids, 28(47), 16455-16462 (2012-11-03)
Lyotropic liquid crystals characterized by a bicontinuous cubic phase (BCP) have a structure characterized by interpenetrated water channels following triply periodic minimal surfaces, which can be stable in excess water conditions and thus suitable in a multitude of applications. The
Woo Song Lee et al.
Bioorganic & medicinal chemistry letters, 15(15), 3573-3575 (2005-06-18)
(R)-Glycerol-monolinoleate 4 and (R)-glycerol-monostearate 5 were isolated from the ethyl acetate extracts of Saururus chinensis roots and (R)- or (S)-fatty acid glycerols 4 and 5 were synthesized for confirming their structures and evaluating their inhibitory activities against Lp-PLA(2). The (R)-4
Nag Jin Choi et al.
Journal of agricultural and food chemistry, 56(22), 10908-10912 (2008-11-01)
This study was designed to isolate bifidobacteria from human intestines that efficiently converts monolinolein, a monoglyceride form of linoleic acid, into conjugated linoleic acid (CLA), as well as to optimize culture conditions for improving CLA production during milk fermentation. Among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service