Skip to Content
Merck

FPCOC5

Flash Point Standard - Cleveland Open Cup

certified reference material, ASTM D 92 (257 °C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

according to ASTM® D92

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

petroleum
pharmaceutical

format

neat

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Cleveland Open Cup Flash Point Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

10 - Combustible liquids

flash_point_f

158.0 °F

flash_point_c

70 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library