Skip to Content
Merck
All Photos(1)

Key Documents

F0120000

Flecainide acetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Flecainide acetate salt, N-(2-Piperidylmethyl)-2,5-bis-(2,2,2-trifluoroethoxy)benzamide acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20F6N2O3 · C2H4O2
CAS Number:
Molecular Weight:
474.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

flecainide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(O)=O.FC(F)(F)COc1ccc(OCC(F)(F)F)c(c1)C(=O)NCC2CCCCN2

InChI

1S/C17H20F6N2O3.C2H4O2/c18-16(19,20)9-27-12-4-5-14(28-10-17(21,22)23)13(7-12)15(26)25-8-11-3-1-2-6-24-11;1-2(3)4/h4-5,7,11,24H,1-3,6,8-10H2,(H,25,26);1H3,(H,3,4)

InChI key

RKXNZRPQSOPPRN-UHFFFAOYSA-N

Gene Information

human ... SCN5A(6331)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Flecainide acetate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Bordier et al.
Annales de cardiologie et d'angeiologie, 45(5), 263-279 (1996-05-01)
Flecainide acetate is a powerful antiarrhythmic initially used to treat ventricular arrhythmias. Despite an excess mortality in this indication and in the presence of heart disease, flecainide remains a good antiarrhythmic for supraventricular tachycardias in a healthy heart.
D Mehta et al.
Cardiovascular drugs and therapy, 1(6), 599-603 (1988-03-01)
Flecainide acetate depresses the rate of depolarization of action potential (Vmax), the so-called "membrane stabilizing action." In the intact heart it has a unique profile of substantial effect on conduction with modest effect on refractoriness. After intravenous administration, clinical electrophysiologic
J C Perry et al.
American heart journal, 124(6), 1614-1621 (1992-12-11)
A review of all published experience with flecainide in infants, children, and fetuses was performed to evaluate the appropriate place of the drug in pediatric practice and to determine dosing guidelines. A total of 704 case references was generated. Flecainide
P Maury et al.
Archives des maladies du coeur et des vaisseaux, 92(2), 273-277 (1999-03-17)
Poisoning with flecainide acetate is rare and associated with a high mortality. This usually occurs after massive ingestion but can also be observed during therapeutic overdose in patients with renal failure or with amiodarone therapy. The prognostic depends on the
S L Chase et al.
Clinical pharmacy, 6(11), 839-850 (1987-11-01)
The chemistry, electrophysiology, pharmacokinetics, clinical use and efficacy, adverse effects, drug interactions, and dosage of encainide hydrochloride and flecainide acetate are reviewed. Encainide and flecainide are class 1c antiarrhythmic agents that slow myocardial conduction and mildly prolong the duration of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service