Skip to Content
Merck
All Photos(1)

Documents

F0040000

Fenbendazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Methyl 5-(phenylthio)-2-benzimidazolecarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O2S
CAS Number:
Molecular Weight:
299.35
Beilstein:
759077
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fenbendazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COC(=O)Nc1nc2cc(Sc3ccccc3)ccc2[nH]1

InChI

1S/C15H13N3O2S/c1-20-15(19)18-14-16-12-8-7-11(9-13(12)17-14)21-10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)

InChI key

HDDSHPAODJUKPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fenbendazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2

Target Organs

Liver,lymph node,Stomach,Nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa O Ezenwa et al.
Science (New York, N.Y.), 347(6218), 175-177 (2015-01-13)
Parasitic worms modulate host immune responses in ways that affect microbial co-infections. For this reason, anthelmintic therapy may be a potent tool for indirectly controlling microbial pathogens. However, the population-level consequences of this type of intervention on co-infecting microbes are
David Villar et al.
Journal of the American Association for Laboratory Animal Science : JAALAS, 46(6), 8-15 (2007-11-13)
This review summarizes findings from toxicologic, carcinogenic, immunologic, and metabolic studies on fenbendazole (FBZ). Currently, FBZ is used to treat or prevent pinworm outbreaks in laboratory rodents. Because antiparasitic treatments usually are not part of experimental designs, interactions from the
Bettencourt P S Capece et al.
Veterinary journal (London, England : 1997), 181(3), 241-250 (2009-01-07)
Albendazole and fenbendazole are methylcarbamate benzimidazole anthelmintics extensively used to control gastrointestinal parasites in domestic animals. These parent compounds are metabolised to albendazole sulfoxide and fenbendazole sulfoxide (oxfendazole), respectively. Both sulfoxide derivatives are anthelmintically active and are manufactured for use
Xiao-Lin Hou et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 962, 20-29 (2014-06-03)
A simple, sensitive and reliable analytical method was developed for the simultaneous determination of 38 veterinary drugs (18 sulfonamides, 11 quinolones and 9 benzimidazoles) and 8 metabolites of benzimidazoles in bovine milk by ultra high performance liquid chromatography-positive electrospray ionization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service