E2642
ExtrAvidin® magnetic beads
For purification of biotinylated macromolecules and complexes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106510
NACRES:
NA.56
Recommended Products
conjugate
magnetic beads
Quality Level
description
for use with mammalian cell and tissue extracts, Non-freezing solution
form
liquid
storage temp.
2-8°C
General description
ExtrAvidin magnetic beads.
Legal Information
ExtrAvidin is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Proteases for biotinylated tag removal for protein purification workflows with related reagents and technical resources.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service