Skip to Content
Merck

D9015

3,3′-Diaminobenzidine tetrahydrochloride hydrate

≥96% purity (TLC), powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H14N4 · 4HCl · xH2O
CAS Number:
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
EC Number:
231-018-9
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

3,3′-Diaminobenzidine tetrahydrochloride hydrate, ISOPAC®

Quality Segment

assay

≥96% (TLC)

form

powder

color

off-white to faint red, to beige

mp

280 °C

solubility

water: 50 mg/mL, clear to slightly hazy

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O.Cl.Cl.Cl.Cl.Nc1ccc(cc1N)-c2ccc(N)c(N)c2

InChI

1S/C12H14N4.4ClH.H2O/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8;;;;;/h1-6H,13-16H2;4*1H;1H2

InChI key

DXWSCXIZIHILNP-UHFFFAOYSA-N

Application

3,3′-Diaminobenzidine tetrahydrochloride hydrate has been used in immunohistological staining procedure and in situ hybridization.

Biochem/physiol Actions

DAB (3,3′-Diaminobenzidine) is utilized in many applications for the visualization of peroxidase activity. In the peroxidase reaction, DAB serves as a hydrogen donor in the presence of peroxide. The oxidized DAB forms an insoluble brown end-product for use in immunohistological and immunoblotting staining procedures.

Packaging

Weight approximate. Packaged in a 100 mL serum bottle with silicon/PTFE septum and aluminum tear seal.

Preparation Note

Dissolving the contents in 100 mL of solvent yields a 0.1% solution.

Legal Information

Isopac is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Injecting a compatible solvent permits preparation of any desired strength solution without exposure.


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library