Skip to Content
Merck

Skip To

D8809

Daunorubicin hydrochloride

meets USP testing specifications

Synonym(s):

Daunomycin hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C27H29NO10 · HCl
CAS Number:
Molecular Weight:
563.98
UNSPSC Code:
41116107
NACRES:
NA.21
PubChem Substance ID:
EC Number:
245-723-4
Beilstein/REAXYS Number:
4229221
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

USP/NF, meets USP testing specifications

form

solid

antibiotic activity spectrum

neoplastics

mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

Cl.COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)C(C)=O

InChI

1S/C27H29NO10.ClH/c1-10-22(30)14(28)7-17(37-10)38-16-9-27(35,11(2)29)8-13-19(16)26(34)21-20(24(13)32)23(31)12-5-4-6-15(36-3)18(12)25(21)33;/h4-6,10,14,16-17,22,30,32,34-35H,7-9,28H2,1-3H3;1H/t10-,14-,16-,17-,22+,27-;/m0./s1

InChI key

GUGHGUXZJWAIAS-QQYBVWGSSA-N

Gene Information

human ... TOP2A(7153)

Biochem/physiol Actions

Naturally fluorescent anthracycline antibiotic, anticancer drug. Substrate for Pgp, MRP-1 and BCRP. Fluorescent property has been exploited for the measurement of drug efflux pump activities as well as resolving the important question of intracellular localization of various multidrug resistance proteins and the role of subcellular organelles (golgi and lysosome) in the sequestration of drugs and its implication in drug resistant phenotypes. Daunorubicin is sequestered into lysosomes according to a pH partitioning type mechanism. Strong inhibitor of DNA and RNA synthesis.
Potent anticancer agent. Inhibits DNA and RNA synthesis as sequence specific ds-DNA intercalating agent.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
30450D0125000PHR3433
description

meets USP testing specifications

description

≥90% (HPLC)

description

European Pharmacopoeia (EP) Reference Standard

description

certified reference material, pharmaceutical secondary standard

mode of action

DNA synthesis | interferes

mode of action

DNA synthesis | interferes

mode of action

-

mode of action

-

antibiotic activity spectrum

neoplastics

antibiotic activity spectrum

neoplastics

antibiotic activity spectrum

-

antibiotic activity spectrum

-

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

300

form

solid

form

powder

form

-

form

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-10 to -25°C

Gene Information

human ... TOP2A(7153)

Gene Information

human ... TOP2A(7153)

Gene Information

human ... TOP2A(7153)

Gene Information

-


Still not finding the right product?

Explore all of our products under Daunorubicin hydrochloride


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Muta. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–5 of 5 Questions  
  1. What factors affect the stability of Product No. D8809, Daunorubicin hydrochloride, in solution?

    1 answer
    1. Per Martindale The Extra Pharmacopoeia, 31st edition, p. 565-3 (1996), stability in solution appeared to be partly related to pH.  Daunorubicin was more stable as the pH of the mixture became more acidic, with the best stability in glucose injection solution (5%) with a pH of 4.5  Although daunorubicin solutions are degraded by light, the effect is reported not to be significant at concentrations of 500 ug/mL or above.  However, below this concentration, precautions should be taken to protect solutions from light, and storage should be in polyethylene or polypropylene containers to minimize adsorptive losses.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. Does Product No. D8809, Daunorubicin hydrochloride, have an expiration date?

    1 answer
    1. Yes, each lot of this product will have an expiration date, and this date (month and year) can be found on the certificate of analysis for the lot.

      Helpful?

  4. Does the assay information on the certificate of analysis for Product No. D8809, Daunorubicin hydrochloride, pertain to a "dry" or "as is" basis?

    1 answer
    1. The assay information on the certificate of analysis is for the product "as is".

      Helpful?

  5. What is this compound soluble in?

    1 answer
    1. Per Martindale The Extra Pharmacopoeia, 31st edition, p. 565-3 (1996), this compound is freely soluble in water and methyl alcohol.  It is slightly soluble in alcohol, very slightly soluble in chloroform, and practically insoluble in acetone.

      Helpful?

Reviews

No rating value

Active Filters