Skip to Content
Merck
All Photos(1)

Documents

D2600000

Diphenhydramine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Diphenylmethoxy-N,N-dimethylethylamine hydrochloride, N-(2-Diphenylmethoxyethyl)-N,N-dimethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO · HCl
CAS Number:
Molecular Weight:
291.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diphenhydramine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].CN(C)CCOC(c1ccccc1)c2ccccc2

InChI

1S/C17H21NO.ClH/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;/h3-12,17H,13-14H2,1-2H3;1H

InChI key

PCHPORCSPXIHLZ-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diphenhydramine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eugene W Schwilke et al.
Journal of forensic sciences, 51(5), 1191-1198 (2006-10-05)
We have previously reported on patterns of drug and alcohol use in fatally injured drivers in Washington State. Here we revisit that population to examine how drug use patterns have changed in the intervening 9 years. Blood and serum specimens
Edward Topp et al.
The Science of the total environment, 439, 136-140 (2012-10-16)
The widely used antihistamine diphenhydramine is present in municipal biosolids, and is detected in runoff from agricultural land fertilized with biosolids. In the present study the kinetics and major pathways of diphenhydramine dissipation in a loam, sandy loam, and clay
A D Rollstin et al.
Clinical toxicology (Philadelphia, Pa.), 51(1), 50-53 (2012-12-01)
There are few reports of acetaminophen overdose in hypothermic patients and even fewer reports describing profound hypothermia. The kinetics, risk of hepatotoxicity, and the possible dose adjustments to N-acetylcysteine (NAC) therapy are not known in this setting. A 37-year-old female
Mohammed Maniruzzaman et al.
International journal of pharmaceutics, 443(1-2), 199-208 (2012-12-25)
The purpose of the study was to investigate and identify the interactions within solid dispersions of cationic drugs and anionic polymers processed by hot-melt extrusion (HME) technique. Propranolol HCl (PRP) and diphenhydramine HCl (DPD) were used as model cationic active
Don W Coulter et al.
Anti-cancer drugs, 24(4), 415-421 (2013-01-19)
A pediatric study has established a maximum tolerated dose (MTD) for temsirolimus (Tem) of more than 150 mg/m intravenously/week. A phase I trial was conducted to establish the MTD for Tem in combination with valproic acid (VPA) in children and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service