Skip to Content
Merck
All Photos(1)

Key Documents

C2285000

Clobetasone butyrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

21-Chloro-9-fluoro-17-hydroxy-16-methylpregna-1,4-diene-3,11,20-trione 17-butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32ClFO5
CAS Number:
Molecular Weight:
478.98
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clobetasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)C(=O)C[C@]12C)C(=O)CCl

InChI

1S/C26H32ClFO5/c1-5-6-22(32)33-26(21(31)14-27)15(2)11-19-18-8-7-16-12-17(29)9-10-23(16,3)25(18,28)20(30)13-24(19,26)4/h9-10,12,15,18-19H,5-8,11,13-14H2,1-4H3/t15-,18-,19-,23-,24-,25-,26-/m0/s1

InChI key

FBRAWBYQGRLCEK-AVVSTMBFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clobetasone butyrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophia Oelzner et al.
Journal of the American Academy of Dermatology, 49(4), 720-722 (2003-09-27)
Chondrodermatitis nodularis chronica helicis is characterized by the presence of small, intensely painful nodules on the free border of the helix of the ear with unknown origin. The lesion is typically localized unilaterally on the right ear. Pressure and trauma
Acute anterior uveitis.
Andreé Curi et al.
Clinical evidence, (9)(9), 691-696 (2004-09-16)
Sweet's syndrome masquerading as figurate erythema.
Barbara Behm et al.
International journal of dermatology, 51(9), 1101-1103 (2012-02-11)
A Parneix-Spake et al.
The Journal of dermatological treatment, 12(4), 191-197 (2002-09-21)
The emollient base of a topical corticosteroid, through its moisturizing properties, can be a useful treatment adjunct. To compare the healing properties of Eumovate trade mark (clobetasone butyrate) 0.05% cream with its emollient base, hydrocortisone 1% cream and with no
E Nivenius et al.
Eye (London, England), 21(7), 968-975 (2006-05-09)
The main objective of this explorative study was to evaluate if tacrolimus ointment could be safer than corticosteroid ointment, with special reference to the intraocular pressure in the treatment of eyelid eczema in patients with atopic keratoconjunctivitis (AKC). Secondary aims

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service