Skip to Content
Merck

Skip To

BP451

Salbutamol

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

α-[(tert-Butylamino)methyl]-4-hydroxy-m-xylene-α,α′-diol, Albuterol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H21NO3
CAS Number:
Molecular Weight:
239.31
EC Number:
242-424-0
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
6405698
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

salbutamol, albuterol

form

crystalline powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(C)NCC(O)c1ccc(O)c(CO)c1

InChI

1S/C13H21NO3/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15/h4-6,12,14-17H,7-8H2,1-3H3

InChI key

NDAUXUAQIAJITI-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Salbutamol BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Biochem/physiol Actions

β2-adrenoceptor agonist

Packaging

Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S8260S01000001012600
Salbutamol British Pharmacopoeia (BP) Reference Standard

BP451

Salbutamol

description

British Pharmacopoeia (BP) Reference Standard

description

-

description

European Pharmacopoeia (EP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

manufacturer/tradename

BP

manufacturer/tradename

-

manufacturer/tradename

EDQM

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

form

crystalline powder

form

-

form

-

form

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-

format

neat

format

-

format

neat

format

neat

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)


Still not finding the right product?

Explore all of our products under Salbutamol


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters