Skip to Content
Merck

Skip To

BP1007

Mannitol

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

D-Mannitol, Mannite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
EC Number:
200-711-8
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
1721898
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

mannitol

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

167-170 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mannitol BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Packaging

Unit quantity: 1200 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.371621.3709663560
Mannitol British Pharmacopoeia (BP) Reference Standard

BP1007

Mannitol

grade

pharmaceutical primary standard

grade

-

grade

-

grade

ACS reagent

manufacturer/tradename

BP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical
semi-solid formulation
solid formulation

application(s)

microbiology

form

powder

form

solid

form

solid

form

powder

format

neat

format

-

format

-

format

-


Still not finding the right product?

Explore all of our products under Mannitol


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters