Skip to Content
Merck
All Photos(1)

Key Documents

B3916

Supelco

Boldine

analytical standard

Synonym(s):

2,9-Dihydroxy-1,10-dimethoxyaporphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO4
CAS Number:
Molecular Weight:
327.37
Beilstein:
94036
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

plant (Peumus boldus molina)

Quality Level

grade

analytical standard

Assay

≥98% (TLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1% isopropanol

mp

157-164  °C

solubility

ethanol: 50 mg/mL

application(s)

food and beverages
forensics and toxicology
veterinary

format

neat

storage temp.

room temp

SMILES string

COc1cc-2c(CC3N(C)CCc4cc(O)c(OC)c-2c34)cc1O

InChI

1S/C19H21NO4/c1-20-5-4-10-7-15(22)19(24-3)18-12-9-16(23-2)14(21)8-11(12)6-13(20)17(10)18/h7-9,13,21-22H,4-6H2,1-3H3/t13-/m0/s1

InChI key

LZJRNLRASBVRRX-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Boldine is an aporphine alkaloid isolated from Boldo tree and other plant species. It is a free radical scavenger with anti-inflammatory activity while exhibiting other pharmacological effects such as antidiabetic, antiplatelet aggregation, antipyretic, antinociceptive, antiatherogenic, hepatoprotective and endothelium-protective activity.

Application

Boldine may be used as an analytical reference standard for the quantification of the analyte in biological samples and pharmaceutical preparations using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Hung et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(13), 3177-3183 (2003-10-30)
In this work we present spectroscopic study of Boldine (aporphine alkaloid) that possesses important biological activities, in particular, in interaction with the promastigotes of Leishmania mexicana. The results show the applicability of autofluorescence of this drug to determinate the possible
C I Cámara et al.
Talanta, 83(2), 623-630 (2010-11-30)
The quantitative determination of boldine alkaloid in boldo leaf extracts by employing cyclic voltammetry, at a liquid/liquid interface as well as the validation of this methodology against the reference method, high performance liquid chromatography (HPLC), are reported in the present
Fiorenza Rancan et al.
Journal of photochemistry and photobiology. B, Biology, 68(2-3), 133-139 (2002-12-07)
Natural substances extracted from lichens and boldo tree were tested in vivo and in vitro as possible UV-light filters. The protection factors were compared with that found for the references: Nivea sun Spray LSF 5, octylmethoxycinnamate (OMC) and 4-tert.-butyl-4'-methoxy dibenzoylmethane
J Fernández et al.
Phytotherapy research : PTR, 23(7), 1024-1027 (2009-01-16)
Peumus boldus Molina (Monimiaceae), commonly referred to as 'boldo', is used in traditional Chilean medicine to treat hepatic and gastrointestinal diseases. Its leaves are rich in antioxidant compounds, principally alkaloids and flavonoids. This study evaluates the protective effect of a
N Santanam et al.
Atherosclerosis, 173(2), 203-210 (2004-04-06)
A corollary to the oxidation hypothesis of atherosclerosis is that the consumption of antioxidants is beneficial. However, the literature is divided in support of this conclusion. In this study, Boldine, an alkaloid of Peumus boldus and reduced form of RU486

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service