Skip to Content
Merck
All Photos(1)

Key Documents

A0496000

Aminoglutethimide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

DL-Aminoglutethimide, 3-(4-Aminophenyl)-3-ethyl-2,6-piperidinedione, 3-(p-Aminophenyl)-3-ethylpiperidine-2,6-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O2
CAS Number:
Molecular Weight:
232.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

aminoglutethimide

manufacturer/tradename

EDQM

mp

152-154 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC1(CCC(=O)NC1=O)c2ccc(N)cc2

InChI

1S/C13H16N2O2/c1-2-13(8-7-11(16)15-12(13)17)9-3-5-10(14)6-4-9/h3-6H,2,7-8,14H2,1H3,(H,15,16,17)

InChI key

ROBVIMPUHSLWNV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Aminoglutethimide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

DL-Aminoglutethimide is a derivative of the sedative glutethimide. Originally introduced as an anticonvulsant, it was found to cause adrenal insufficiency. Blocks adrenal steroidogenesis by inhibiting the enzymatic conversion of cholesterol to pregnenolone. It also blocks the peripheral conversion (aromatization) of androgenic precursors to estrogens. The D-isomer is 30 times more potent at inhibiting aromatase activity, whereas the L-isomer is more potent at inhibiting cholesterol side-chain cleavage (steroidogenesis).

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P E Lønning et al.
Drugs, 35(6), 685-710 (1988-06-01)
During the last decade aminoglutethimide has been recognised as a valuable alternative in endocrine therapy for advanced breast cancer. Although some side effects do occur, most often these are initial effects which subside within a few weeks, and cessation of
R J Santen
Breast cancer research and treatment, 1(3), 183-202 (1981-01-01)
Fifty to sixty percent of postmenopausal women with estrogen receptor positive metastatic breast cancer respond objectively to surgical ablation of the pituitary or adrenal glands. Several investigators have recently developed medical alternatives to surgical ablative therapy for these patients. This
Aminoglutethimide in the treatment of advanced breast cancer.
R C Stuart-Harris et al.
Cancer treatment reviews, 11(3), 189-204 (1984-09-01)
A M Brodie et al.
Critical reviews in oncology/hematology, 5(4), 361-396 (1986-01-01)
Approximately one third of human breast carcinomas are hormone dependent and regress upon reduction of circulating estrogen levels. Traditional treatment strategies utilized surgical ablative methods to lower estrogen concentrations as treatment of breast cancer. Currently, investigative emphasis is focused upon
Aminoglutethimide: a "side-effect" turned to therapeutic advantage.
S W Hughss et al.
Postgraduate medical journal, 46(537), 409-416 (1970-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service