Skip to Content
Merck
All Photos(1)

Key Documents

A0220000

Aciclovir

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Acycloguanosine, 9-[(2-Hydroxyethoxy)methyl]guanine, Acyclovir

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N5O3
CAS Number:
Molecular Weight:
225.20
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

acyclovir

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NC1=Nc2c(ncn2COCCO)C(=O)N1

InChI

1S/C8H11N5O3/c9-8-11-6-5(7(15)12-8)10-3-13(6)4-16-2-1-14/h3,14H,1-2,4H2,(H3,9,11,12,15)

InChI key

MKUXAQIIEYXACX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Aciclovir EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Daniel S Owers et al.
The Cochrane database of systematic reviews, 2(2), CD005133-CD005133 (2013-03-02)
Cytomegalovirus (CMV) is a significant cause of morbidity and mortality in solid organ transplant recipients. Pre-emptive treatment of patients with CMV viraemia using antiviral agents has been suggested as an alternative to routine prophylaxis to prevent CMV disease. This is
Adalbert Krawczyk et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(17), 6760-6765 (2013-04-10)
Despite the availability of antiviral chemotherapy, herpes simplex virus type 1 (HSV-1) and type 2 (HSV-2) infections remain a severe global health problem. Of particular concern is the growing incidence of drug resistance in immunocompromised patients, which stresses the urgency
Monique van Velzen et al.
The Journal of infectious diseases, 208(9), 1359-1365 (2013-08-01)
Long-term acyclovir (ACV) prophylaxis, recommended to prevent recurrent herpes simplex virus type 1 (HSV-1) ocular disorders, may pose a risk for ACV-refractory disease due to ACV resistance. We determined the effect of ACV prophylaxis on the prevalence of corneal ACV-resistant
Sonia Burrel et al.
Journal of clinical virology : the official publication of the Pan American Society for Clinical Virology, 58(1), 321-324 (2013-06-05)
Herpes simplex virus type 1 (HSV-1) is a leading cause of corneal blindness. Acyclovir (ACV) constitutes the standard treatment of HSV infections including herpetic keratitis (HK). HSV resistance to ACV is mainly described in immunocompromised patients. We describe two cases
Swetha G Pinninti et al.
Pediatric clinics of North America, 60(2), 351-365 (2013-03-14)
Neonatal herpes simplex virus infections are uncommon, but because of the morbidity and mortality associated with the infection they are often considered in the differential diagnosis of ill neonates. The use of polymerase chain reaction for diagnosis of central nervous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service