Skip to Content

95039

M-Gredient® Casein Peptone for Biotechnology and Microbiology

Pancreatically Digested

Synonym(s):

Peptone from casein, Peptone from casein, tryptic digest, Tryptone enzymatic digest from casein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
EC Number:
293-428-4
NACRES:
NA.85
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

M-Gredient® Casein Peptone B1, Pancreatically Digested

biological source

bovine milk

Quality Segment

form

powder

composition

amino-nitrogen, ≥3.5% , Total Nitrogen (N), ≥12%

packaging

pkg of 1 kg, pkg of 5 kg

ign. residue

≤17.0%

loss

≤6.0% loss on drying

pH

6.0-7.5

solubility

H2O: 2%, clear, faintly to slightly yellow

application(s)

food and beverages
(Microbiology and Biotechnology)

storage temp.

15-25°C

General description

Tryptone is a general-purpose peptone produced by the digestion of casein with the protease trypsin. It is used for the cultivation of most bacteria, fungi, and some protozoa. Casein is the main protein of milk, and a rich source of nitrogenous nutrients (in the amino acids). This peptone is recommended for culture media in analytical microbiology and in industrial fermentation.

Application

Tryptone from enzymatic casein digest can be developed for use in molecular genetic media such as – NZCYM, NZYM and NZM broth. The enriched NZ media can be used to: -
  • Grow recombinant strains of E. coli
  • Propagate theta bacteriophages

Legal Information

M-GREDIENT is a registered trademark of Merch KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under M-Gredient® Casein Peptone B1


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library