Skip to Content
Merck
All Photos(1)

Documents

92330

Sigma-Aldrich

Trimethyl borate

purum, ≥99.0% (GC)

Synonym(s):

Boric acid trimethyl ester, Methyl borate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
B(OCH3)3
CAS Number:
Molecular Weight:
103.91
Beilstein:
1697939
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.59 (vs air)

Quality Level

grade

purum

Assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.346 (lit.)
n20/D 1.358

bp

68-69 °C (lit.)

mp

−34 °C (lit.)

density

0.932 g/mL at 20 °C (lit.)

SMILES string

COB(OC)OC

InChI

1S/C3H9BO3/c1-5-4(6-2)7-3/h1-3H3

InChI key

WRECIMRULFAWHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethyl borate (TMB) can be used:
  • For the synthesis of luminogens having triphenylamine core and tetraphenylethene peripheral moieties.
  • For the synthesis of ammonia borane and trialkylamine boranes.
  • In the reduction of carboxylic acids in the presence of borane-methyl sulfide.
  • As a reagent to crosslink phopshate complexes.
  • As a source of boron to synthesize boron nitride nanotubes by chemical vapor deposition (CVD) method.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - (own results)

Flash Point(C)

-11 °C - (own results)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic synthesis using borane-methyl sulfide. II. Reduction of aromatic carboxylic acids in the presence of trimethyl borate.
Lane CF, et al.
The Journal of Organic Chemistry, 39(20), 3052-3054 (1974)
Changing the Behavior of Chromophores from Aggregation?Caused Quenching to Aggregation?Induced Emission: Development of Highly Efficient Light Emitters in the Solid State.
Yuan W Z, et al.
Advanced Materials, 22(19), 2159-2163 (2010)
Thermal-heating CVD synthesis of BN nanotubes from trimethyl borate and nitrogen gas.
Lin FH, et al.
Materials Chemistry and Physics, 107(1), 115-121 (2008)
One-Pot Synthesis of Ammonia-Borane and Trialkylamine-Boranes from Trimethyl Borate.
Veeraraghavan RP, et al.
Organic Letters, 14(24), 6119-6121 (2012)
R Meder et al.
Solid state nuclear magnetic resonance, 15(1), 69-72 (2000-07-21)
Boron-11 nuclear magnetic resonance imaging and spectroscopy have been used to characterise the nature and distribution of boron compounds after preservative treatment of radiata pine wood with trimethylborate (TMB). One day after treatment, 11B magnetic resonance imaging microscopy showed significant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service