Skip to Content
Merck
All Photos(1)

Documents

91521

Supelco

Decanoyl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-1-propanaminium inner salt, L-Carnitine decanoyl ester, C10-Carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H33NO4
CAS Number:
Molecular Weight:
315.45
Beilstein:
4145305
UNSPSC Code:
41116107
NACRES:
NB.21

grade

analytical standard

Quality Level

Assay

≥94.0% (HPLC)

optical activity

[α]/D -15±3°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

impurities

≤10% water

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCCCCCCC)=O)CC([O-])=O

InChI

1S/C17H33NO4/c1-5-6-7-8-9-10-11-12-17(21)22-15(13-16(19)20)14-18(2,3)4/h15H,5-14H2,1-4H3/t15-/m1/s1

InChI key

LZOSYCMHQXPBFU-OAHLLOKOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decanoyl-L-carnitine, belonging to the class of acylcarnitines, serves as a specific diagnostic metabolite biomarker, found in medium-chain acyl CoA dehydrogenase (MCAD) deficiency especially in newborns.

Application

Decanoyl-L-carnitine may be used as an analytical reference standard for the quantification of the analyte in the following:
  • In dried blood spots using chip-based nanoelectrospray source coupled with high resolution mass spectrometry (chip-based nanoESI- HRMS) and targeted liquid chromatography-tandem mass spectrometry (LC/MS/MS).
  • In biological samples using high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC/MS/MS).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supervised machine learning techniques for the classification of metabolic disorders in newborns
Baumgartner C, et al.
Bioinformatics, 20(17), 2985-2996 (2004)
Quantification of metabolites in dried blood spots by direct infusion high resolution mass spectrometry
Van der Ham M, et al., et al.
Analytica Chimica Acta, 979, 45-50 (2017)
Non-invasive test using palmitate in patients with suspected fatty acid oxidation defects: disease-specific acylcarnitine patterns can help to establish the diagnosis
Janzen N, et al.
Orphanet Journal of Rare Diseases, 12(1), 187-187 (2017)
Differences between acylcarnitine profiles in plasma and bloodspots
Diekman FE, et al.
Molecular Genetics and Metabolism, 110(1-2), 116-121 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service