Skip to Content
Merck

90243

Multielement standard solution 1 for ICP

TraceCERT®, in 10% nitric acid

Synonym(s):

ICP Multi-Element Calibration Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

quality

in 10% nitric acid

composition

Ag, 10 mg/L , Al, 50 mg/L , B, 50 mg/L , Ba, 10 mg/L , Bi, 100 mg/L , Ca, 10 mg/L , Cd, 10 mg/L , Co, 10 mg/L , Cr, 50 mg/L , Cu, 10 mg/L , Fe, 10 mg/L , K, 100 mg/L , Li, 50 mg/L , Mg, 10 mg/L , Mn, 10 mg/L , Mo, 50 mg/L , Na, 50 mg/L , Ni, 50 mg/L , Pb, 100 mg/L , Sr, 10 mg/L , Tl, 50 mg/L , Zn, 10 mg/L

concentration

in 10% HNO3

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

multi-component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Search & download your certificate here.

Application

Multielement standard solution 1 has been used in the analysis of pollutants like trace elements in water using total reflection x-ray fluorescence (TXRF) method and inductively coupled plasma mass spectrometry(ICP-MS).

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website
For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

supp_hazards

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

WGK 3

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library