Skip to Content
Merck
All Photos(1)

Key Documents

900690

Sigma-Aldrich

N,N-Diisopropylethylamine

purified by redistillation, ZerO2®, 99.5%

Synonym(s):

N-Ethyldiisopropylamine, DIPEA, Ethyldiisopropylamine, ‘Hünig’s base’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NC2H5
CAS Number:
Molecular Weight:
129.24
EC Number:
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21
Pricing and availability is not currently available.

vapor pressure

31 mmHg ( 37.7 °C)

Quality Level

Assay

99.5%

form

liquid

refractive index

n20/D 1.414 (lit.)

bp

127 °C (lit.)

mp

<−50 °C (lit.)

density

0.742 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCN(C(C)C)C(C)C

InChI

1S/C8H19N/c1-6-9(7(2)3)8(4)5/h7-8H,6H2,1-5H3

InChI key

JGFZNNIVVJXRND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Diisopropylethylamine, also known as Hünig′s base, is a sterically hindered amine. It is a non-nucleophilic base commonly employed for substitution reactions.

Application

N,N-Diisopropylethylamine may be used in the synthesis of mannosylated ovalbumin peptides.[1]
Proton scavenger used in peptide coupling[2], enolboration[3], Pd(0)-catalyzed alkoxycarbonylation of allyl phosphates and acetates[4], and as a catalyst in vinyl sulfone synthesis.[5]

Features and Benefits

ZerO2® products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2® is a trademark of Sigma-Aldrich Co. LLC
ZerO2 is a registered trademark of Merck KGaA, Darmstadt, Germany
ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.1 °F

Flash Point(C)

9.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 58, 7162-7162 (1993)
Synthetic Communications, 23, 3073-3073 (1993)
The Journal of Organic Chemistry, 59, 695-695 (1994)
The Journal of Organic Chemistry, 58, 1538-1538 (1993)
Pavla Simerska et al.
Drug delivery and translational research, 4(3), 246-255 (2015-03-20)
Peptide-based vaccine delivery can be hampered by rapid peptidase activity and poor inherent immunogenicity. The self-adjuvanting lipid core peptide system (LCP) has been shown to confer improved stability and immunogenicity on peptide epitopes of group A Streptococcus, Chlamydia, hookworm, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service