Skip to Content
Merck
All Photos(2)

Key Documents

861375

Sigma-Aldrich

Wright stain

certified by the Biological Stain Commission

Synonym(s):

Eosin Methylene blue according to Wright

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

Assay

≤4% (HPLC)

form

powder

concentration

≤4% (HPLC)

solubility

methanol: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[s]5c6c(nc7c5cc(cc7)N(C)C)cc[c](c6)=[N+](C)C.Brc1c2[o+]c3c(c(c2cc(c1O)Br)c4c(cccc4)C(=O)O)cc(c(c3Br)O)Br

InChI

1S/C20H8Br4O5.C16H18N3S/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13/h1-6H,(H2-,25,26,27,28);5-10H,1-4H3/q;+1/p+1

InChI key

AXIKDPDWFVPGOD-UHFFFAOYSA-O

Looking for similar products? Visit Product Comparison Guide

General description

Wright Stain, a modified Romanowsky stain, is intended for differential staining of the cellular elements of blood. It comprises polychrome methylene blue and eosin B. Upon staining blood films with Wright Stain, the white blood cell nucleus and cytoplasm display characteristic blue or pink hues.

Application

  • Wright Stain is intended for use in staining peripheral blood smears and bone marrow smears.
  • It has been used to study the cytogenetics of human hematopoietic cells.
  • It has also been employed in the karyotyping of human embryonic stem cells.
  • It is also used for G-banding and helps in the analysis of poorly spread metaphases.
Reported to be superior to May-Grunwald Stain in blood staining.

Features and Benefits

  • The purified dyes in Wright Stain eliminate inconsistent staining and yield reproducible lot-to-lot chromogenic responses.
  • Reported to be superior to May-Grunwald Stain in blood staining.
  • Certified by the Biological Stain Commission.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Generation of FHL2 homozygous knockout lines from human embryonic stem cells by CRISPR/Cas9-mediated ablation
Chang, et al.
Stem Cell Research, 27 (2018)
Conn?s Biological Stains: A Handbook of Dyes, Stains and Fluorochromes for Use in Biology and Medicine (10th ed.)
RW Horobin, JA Kiernan
Conn?s Biological Stains (2002)
Chromosome Analysis Protocols (1994)
Phenotypic, Functional, and Safety Control at Preimplantation Phase of MSC-Based Therapy.
Lech W, et al.
Stem Cells International, 2016, 2514917-2514917 (2016)
Genetic Inactivation of CD33 in Hematopoietic Stem Cells to Enable CAR T Cell Immunotherapy for Acute Myeloid Leukemia
Kim, et al.
Cell, 173(6) (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service