Skip to Content
Merck
All Photos(1)

Key Documents

80030

Supelco

Bis(2-ethylhexyl) phthalate

Selectophore

Synonym(s):

DEHP, Phthalic acid bis(2-ethylhexyl ester), ‘Dioctyl’ phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
Beilstein:
1890696
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
grade:
for ion-selective electrodes
bp:
386 °C (lit.)

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

viscous liquid (clear)

bp

386 °C (lit.)

density

0.985 g/mL at 20 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC

InChI

1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3

InChI key

BJQHLKABXJIVAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2-ethylhexyl) phthalate (DEHP), is a phthalate ester which is a dialkyl or aryl/alkyl diesters of phthalic acid. It is mostly used as additives for imparting flexibility to polymers in plastics technology. Generally, phthalate ester are hydrophobic compounds which displays strong potential for adsorption.
Visit our Sensor Applications portal to learn more.

Application

DEHP may be used as plasticisers in studying the transcriptional activity of human sodium/iodide symporter.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

392.0 °F - Pensky-Martens closed cup

Flash Point(C)

200 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oral toxicity of bis(2-ethylhexyl) phthalate during pregnancy and suckling in the Long-Evans rat.
Arcadi FA
Food And Chemical Toxicology, 36(11), 963-970 (1998)
Removal of bis(2-ethylhexyl) phthalate at a sewage treatment plant.
Marttinen SK
Water Research, 37(6), 1385-1393 (2003)
Océane Albert et al.
Toxicological sciences : an official journal of the Society of Toxicology, 164(1), 129-141 (2018-06-27)
Di(2-ethylhexyl) phthalate (DEHP), a widely used plasticizer, is a ubiquitous environmental contaminant and may act as an endocrine disruptor. Early life exposures to DEHP may result in anti-androgenic effects, impairing the development of the male reproductive tract. However, data on
Human biological monitoring of diisononyl phthalate and diisodecyl phthalate: a review.
Saravanabhavan G, Murray J.
Journal of Environmental and Public Health, 1-11 (2012)
The promoter of the human sodium/iodide symporter responds to certain phthalate plasticisers.
Breous E, Wenzel A, Loos U.
Molecular and Cellular Endocrinology, 244(1-2), 75-78 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service