Skip to Content
Merck
All Photos(1)

Key Documents

78610

Sigma-Aldrich

Phenylglyoxylic acid

purum, ≥98.0% (T)

Synonym(s):

α-Oxophenylacetic acid, Benzoylformic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCOOH
CAS Number:
Molecular Weight:
150.13
Beilstein:
606718
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

purum

Quality Level

Assay

≥98.0% (T)

form

crystals

bp

84 °C/0.1 mmHg

mp

62-65 °C (lit.)
63-67 °C

solubility

methanol: 0.1 g/mL, clear

functional group

carboxylic acid
ketone
phenyl

SMILES string

OC(=O)C(=O)c1ccccc1

InChI

1S/C8H6O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5H,(H,10,11)

InChI key

FAQJJMHZNSSFSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caleb J Allpress et al.
Journal of the American Chemical Society, 135(2), 659-668 (2012-12-12)
Mononuclear Fe(II) complexes ([(6-Ph(2)TPA)Fe(PhC(O)C(R)C(O)Ph)]X (3-X: R = OH, X = ClO(4) or OTf; 4: R = H, X = ClO(4))) supported by the 6-Ph(2)TPA chelate ligand (6-Ph(2)TPA = N,N-bis((6-phenyl-2-pyridyl)methyl)-N-(2-pyridylmethyl)amine) and containing a β-diketonate ligand bound via a six-membered chelate ring
Sébastien Gagné
Rapid communications in mass spectrometry : RCM, 26(7), 845-852 (2012-03-01)
Ultra-performance liquid chromatography coupled with tandem mass spectrometry was used for the biological monitoring of workers occupationally exposed to solvents. The method was developed using a triple quadrupole to investigate the relevant urinary metabolites of styrene, namely mandelic acid and
Sirilak Wongvijitsuk et al.
International journal of hygiene and environmental health, 214(2), 127-137 (2010-10-30)
The present study aimed to evaluate the effects of styrene exposure at levels below the recommended standards of the Threshold Limit Value (TLV-TWA(8)) of 20 ppm (ACGIH, 2004) in reinforced-fiberglass plastics workers. Study subjects comprised 50 exposed workers and 40
Zhi-de Zhou et al.
International journal of biological macromolecules, 47(1), 21-26 (2010-04-20)
Saccharomyces cerevisiae alcohol dehydrogenase (SCAD) was effectively immobilized on hybrid alginate-chitosan beads which are hardened with glutaraldehyde. Immobilization conditions and characterization of the immobilized enzyme were investigated. Orthogonal test design and intuitive analysis method were employed to evaluate the effects
Gerhard Triebig et al.
International archives of occupational and environmental health, 82(4), 463-480 (2008-09-03)
Associations between occupational styrene exposure and impairment of hearing function were investigated, guided by three questions: are there hearing losses concerning high frequency and standard audiometric test? Are there dose-response relationships and measurable thresholds of effects? Are there signs of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service