Skip to Content
Merck
All Photos(1)

Key Documents

76929

Sigma-Aldrich

1-Pentanol

puriss. p.a., ACS reagent, ≥99.0% (GC)

Synonym(s):

n-Amyl alcohol, Pentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730975
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
136-138 °C (lit.)
vapor pressure:
1 mmHg ( 13.6 °C)
1.5 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

3 (vs air)

vapor pressure

1 mmHg ( 13.6 °C)
1.5 mmHg ( 20 °C)

Assay

≥99.0% (GC)

form

liquid

expl. lim.

10 %, 100 °F

technique(s)

GC/GC: suitable

impurities

substances darkened by H2SO4, in accordance
≤0.005% free acid (as CH3COOH)
≤0.1% carbonyl compounds (as HCHO)
≤0.1% water

evapn. residue

≤0.002%

color

clear

refractive index

n20/D 1.409 (lit.)
n20/D 1.410

bp

136-138 °C (lit.)

mp

−78 °C (lit.)

density

0.811 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.5 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.5 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CCCCCO

functional group

hydroxyl

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

AMQJEAYHLZJPGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Pentanol is a linear alcohol. Pentane, a ″next generation″ alcohol fuel,[1] is a promising substitute of conventional gasoline and diesel fuels. Its combustion characteristics have been investigated.[2] Kinetics of thermal decomposition of its isomeric forms (1-pentanol, 2-methyl-1-butanol, and 3-methyl-1-butanol) have been reported.[1]

Application

1-Pentanol may be used as a cosurfactant in the water-in-oil mixed surfactant microemulsions.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long Zhao et al.
The journal of physical chemistry. A, 116(37), 9238-9244 (2012-08-23)
Pentanol is one of the promising "next generation" alcohol fuels with high energy density and low hygroscopicity. In the present work, dominant reaction channels of thermal decomposition of three isomers of pentanol: 1-pentanol, 2-methyl-1-butanol, and 3-methyl-1-butanol were investigated by CBS-QB3
Kaushik Kundu et al.
Journal of colloid and interface science, 385(1), 96-110 (2012-08-14)
The present study is focused on the evaluation of the interfacial composition, thermodynamic properties, and structural parameters of water-in-oil mixed surfactant microemulsions [(cetylpyridinium chloride, CPC+polyoxyethylene (20) cetyl ether, Brij-58 or polyoxyethylene (20) stearyl ether, Brij-78)/1-pentanol/n-heptane, or n-decane] under various physicochemical
Acid/base- and anion-controllable organogels formed from a urea-based molecular switch.
Sheng-Yao Hsueh et al.
Angewandte Chemie (International ed. in English), 49(48), 9170-9173 (2010-10-19)
Experimental and detailed kinetic modeling study of 1-pentanol oxidation in a JSR and combustion in a bomb.
Togbe C, et al.
Proceedings of the Combustion Institute, 33(1), 367-374 (2011)
Gizelle Cristina Bedendo et al.
Journal of chromatography. A, 1217(42), 6449-6454 (2010-09-14)
A sensitive and precise analysis using hollow fiber renewal liquid membrane (HFRLM) extraction followed by high performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) is described for determination of five sulfonamides in honey samples. In this procedure, the organic solvent introduced directly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service