Skip to Content
Merck
All Photos(2)

Key Documents

76706

Supelco

Tryptamine

analytical standard

Synonym(s):

2-(3-Indolyl)ethylamine, 3-(2-Aminoethyl)indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2
CAS Number:
Molecular Weight:
160.22
Beilstein:
125513
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (GC)
97.5-102.5% (NT)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

137 °C/0.15 mmHg (lit.)

mp

113-116 °C (lit.)

suitability

conforms to structure for Infrared spectrum

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

NCCc1c[nH]c2ccccc12

InChI

1S/C10H12N2/c11-6-5-8-7-12-10-4-2-1-3-9(8)10/h1-4,7,12H,5-6,11H2

InChI key

APJYDQYYACXCRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tryptamines which are usually found in plants, fungi, animals, etc. are categorized under the monoamine alkaloids class of compounds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tryptamine has been used as a reference standard for the determination of tryptamine in fish and fish products by liquid chromatography (LC) combined with single-stage Orbitrap high resolution mass spectrometry (HRMS), in Polish wine samples by direct solid phase microextraction coupled to gas chromatography-mass spectrometry (DI-SPME-GC-MS), in Korean turbid rice wine, Makgeolli by ultra-high-pressure liquid chromatography-electrospray tandem mass spectrometry (UHPLC-ESI-MS/MS) and in beer by a non-woven nanofibre mat in combination with solid surface-room temperature phosphorescence (SS-RTP).

Biochem/physiol Actions

Vasoactive; may have a neuromodulator function; biogenic amine formed from the decarboxylation of tryptophan by L-aromatic amino acid decarboxylase.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

365.0 °F - closed cup

Flash Point(C)

185 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct solid phase microextraction combined with gas chromatography-Mass spectrometry for the determination of biogenic amines in wine.
Papageorgiou M, et al.
Talanta, 183(2), 276-282 (2018)
Recreational use, analysis and toxicity of tryptamines.
Tittarelli R, et al.
Current Neuropharmacology, 13(1), 26-46 (2015)
Easy and Fast Method for the Determination of Biogenic Amines in Fish and Fish Products with Liquid Chromatography Coupled to Orbitrap Tandem Mass Spectrometry.
Kaufmann A and Maden K
Journal of AOAC (Association of Official Analytical Chemists) International, 101(2), 336-341 (2018)
The identification and quantification of biogenic amines in Korean turbid rice wine, Makgeolli by HPLC with mass spectrometry detection.
Lee S, et al.
LWT--Food Science and Technology, 62(1), 350-356 (2015)
Novel optical sensing film based on a functional nonwoven nanofibre mat for an easy, fast and highly selective and sensitive detection of tryptamine in beer.
Ramon-Marquez T, et al.
Biosensors And Bioelectronics, 79(1), 600-607 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service