Skip to Content
Merck
All Photos(1)

Documents

75563

Supelco

Ethyl butyrate

analytical standard

Synonym(s):

Butyric acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C(O)OC2H5
CAS Number:
Molecular Weight:
116.16
Beilstein:
506331
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4 (vs air)

vapor pressure

15.5 mmHg ( 25 °C)

Assay

≥99.5% (GC)

autoignition temp.

865 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.392 (lit.)
n20/D 1.393

bp

120 °C (lit.)

mp

−93 °C (lit.)

density

0.875 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCC(=O)OCC

InChI

1S/C6H12O2/c1-3-5-6(7)8-4-2/h3-5H2,1-2H3

InChI key

OBNCKNCVKJNDBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl butyrate belongs to the class of esters and is generally employed as a flavor enhancer in commercial food products.

Application

Ethyl butyrate may be used as an analytical reference standard for the quantification of the analyte in orange essence oil samples and has been used as a reference standard for the determination of the analyte in dry and sweet white wines using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of aroma active compounds in orange essence oil using gas chromatography?olfactometry and gas chromatography?mass spectrometry
Hognadottir A and Rouseff.LR
Journal of Chromatography A, 998(1-2), 201-211 (2003)
Determination of esters in dry and sweet white wines by headspace solid-phase microextraction and gas chromatography
Rodriguez-Bencomo JJ, et al.
Journal of Chromatography A, 963(1-2), 213-223 (2002)
Citrus Essential Oils: Flavor and Fragrance (2011)
Boriana Atanasova et al.
Chemical senses, 30(3), 209-217 (2005-03-03)
The qualitative perceptual interactions in three binary mixtures of wine odorants were studied: isoamyl acetate (fruity note)/whisky lactone (woody note), ethyl butyrate (fruity note)/whisky lactone (woody note) and ethyl butyrate (fruity note)/guaiacol (woody note). For each binary mixture, the perceived
D Labbe et al.
Chemical senses, 32(3), 205-214 (2006-11-03)
The impact of olfactory perception on sweetness was explored in a model solution using odorants at subthreshold concentrations. First, the impact of 6 odorants, previously described in the literature as congruent with sweetness, was investigated at suprathreshold level in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service