Skip to Content
Merck
All Photos(1)

Key Documents

73822

Supelco

Genistin

analytical standard

Synonym(s):

4′,5,7-Trihydroxyisoflavone 7-glucoside, Genistein 7-glucoside, Genistein-7-O-β-D-glucopyranoside, Genistoside, NSC 5112

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Beilstein:
64479
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](Oc2cc(O)c3C(=O)C(=COc3c2)c4ccc(O)cc4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O10/c22-7-15-18(26)19(27)20(28)21(31-15)30-11-5-13(24)16-14(6-11)29-8-12(17(16)25)9-1-3-10(23)4-2-9/h1-6,8,15,18-24,26-28H,7H2/t15-,18-,19+,20-,21-/m1/s1

InChI key

ZCOLJUOHXJRHDI-CMWLGVBASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Genistin belongs to the class of isoflavone glycosides generally extracted from soybeans.

Application

Genistin has been used as an analytical reference standard for the analysis of genistin in:
  • Legumes by ultra-high performance liquid chromatography coupled to tandem mass spectrometry (UHPLC-MS/MS) with electrospray ionization (ESI) source and multiple reaction monitoring (MRM) detection.

It may be used as an analytical reference standard for the determination of genistin in:
  • Soy-based foods and dietary supplements by LC coupled to absorbance detection and isotope-dilution mass spectrometry (IDMS), equipped with ESI source and selected ion monitoring (SIM) mode.
  • Soy products by colloidal gold-based indirect competitive immunochromatographic assay (ICA).
  • Rainbow trout muscle by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction and LC-ESI-MS/MS.
  • Soybean flour by matrix solid phase dispersion extraction (MSPD) and LC with ultraviolet-diode array detection (UV-DAD).
  • Soymilk samples by HPLC-UV operating under the gradient elution mode.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Selective inhibitor of mammalian terminal deoxynucleotidyl transferase (TdT), with no measurable effect on mammalian or microbial DNA polymerases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: glycyrrhiza

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Colloidal gold-based indirect competitive immunochromatographic assay for rapid detection of bioactive isoflavone glycosides daidzin and genistin in soy products.
Sakamoto S, et al.
Food Chemistry, 194, 191-195 (2016)
Effective clean-up and ultra high-performance liquid chromatography-tandem mass spectrometry for isoflavone determination in legumes.
Vila-Donat P, et al.
Food Chemistry, 174, 487-494 (2015)
Analysis of water-soluble bioactive compounds in commonly consumed soymilk in China.
Li YR, et al.
J. Food Compos. Anal., 46(4), 29-35 (2016)
Isoflavones quantification in rainbow trout muscle by QuEChERS tecnique and liquid chromatography coupled with mass spectrometry.
Merlanti R, et al.
J. Food Compos. Anal., 70(4), 114-124 (2018)
Determination of Isoflavones in Soybean Flour by Matrix Solid-Phase Dispersion Extraction and Liquid Chromatography with UV-Diode Array Detection.
Aresta A, et al.
Journal of Food Quality, 2017(4), 114-124 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service