Skip to Content
Merck
All Photos(1)

Key Documents

73148

Supelco

3-Nitrobenzyl alcohol

suitable for mass spectrometry (MS), ≥99.5%

Synonym(s):

NOBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2OH
CAS Number:
Molecular Weight:
153.14
Beilstein:
2044769
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.5% (GC)
≥99.5%

form

(Powder or Crystals or Solid or Liquid)

technique(s)

H-NMR: suitable
mass spectrometry (MS): suitable

bp

175-180 °C/3 mmHg (lit.)

mp

26-32 °C (lit.)

density

1.29 g/mL at 20 °C (lit.)

SMILES string

OCc1cccc(c1)[N+]([O-])=O

InChI

1S/C7H7NO3/c9-5-6-2-1-3-7(4-6)8(10)11/h1-4,9H,5H2

InChI key

CWNPOQFCIIFQDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Nitrobenzyl alcohol (3-NBA) is most commonly used matrix for spectrometry methods. It helps in the analysis of porphyrins, cyclic peptides, alkali metal cationized fatty acids for collisional activation and also certain organoruthenium compounds.

Application

3-NBA has been used as liquid matrix in Ionization in liquid secondary ion mass spectrometry.
Matrix used in fast atom bombardment mass spectrometrycitationcitationcitation

Other Notes

Matrix used in fast atom bombardment mass spectrometry

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T.W. Chan et al.
Org. Mass Spectrom., 27, 53-53 (1992)
J. Meili et al.
Org. Mass Spectrom., 19, 581-581 (1984)
S Hartmann et al.
Biological mass spectrometry, 23(6), 362-368 (1994-06-01)
Fast atom bombardment mass spectra were successfully recorded for intact glycosylphenolphthiocerol dimycocerosates (phenolic glycolipids, PGLs) from Mycobacterium kansasii, M. leprae, M. tuberculosis, M. marinum, M. bovis and M. haemophilum. Characteristic fragment ions from the loss of the oligosaccharide moiety and
Harry J Sterling et al.
Journal of the American Society for Mass Spectrometry, 23(2), 191-200 (2011-12-14)
The effects of aqueous solution supercharging on the solution- and gas-phase structures of two protein complexes were investigated using traveling-wave ion mobility-mass spectrometry (TWIMS-MS). Low initial concentrations of m-nitrobenzyl alcohol (m-NBA) in the electrospray ionization (ESI) solution can effectively increase
3-Nitrobenzyl alcohol has wide applicability as a matrix for FAB-MS.
Sweetman, Brian J., and Ian A. Blair.
Biological Mass Spectrometry, 17.4, 337-340 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service