Skip to Content
Merck
All Photos(2)

Documents

72609

Supelco

Coumarin

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1,2-Benzopyrone, 1-Benzopyran-2-one, 2H-Chromen-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
Beilstein:
383644
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor pressure

0.01 mmHg ( 47 °C)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

298 °C (lit.)

mp

68-73 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

O=C1Oc2ccccc2C=C1

InChI

1S/C9H6O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H

InChI key

ZYGHJZDHTFUPRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: cinnamomum curcuma hedera humulus melissa mentha salvia sambucus zingiber

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

323.6 °F - closed cup

Flash Point(C)

162 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B G Lake
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 37(4), 423-453 (1999-07-27)
The metabolism, toxicity and results of tests for carcinogenicity have been reviewed with respect to the safety for humans of coumarin present in foodstuffs and from fragrance use in cosmetic products. Coumarin is a natural product which exhibits marked species
Shigehiro Sumiya et al.
The journal of physical chemistry. A, 117(7), 1474-1482 (2013-01-25)
A coumarin-amide-dipicolylamine linkage (L) was synthesized and used as a fluorescent receptor for metal cations in water. The receptor dissolved in water with neutral pH shows almost no fluorescence due to the photoinduced electron transfer (PET) from the amide and
Shota Morimoto et al.
Chemical communications (Cambridge, England), 49(18), 1811-1813 (2013-01-26)
A photo-switchable fluorescent flagging approach has been developed to identify photoaffinity-labeled peptides in target protein. Upon photochemical release of the ligand, the protein was newly modified with a coumarin in place of the previously attached biotin. It allowed us to
Klaus Abraham et al.
Molecular nutrition & food research, 54(2), 228-239 (2009-12-22)
Coumarin is a secondary phytochemical with hepatotoxic and carcinogenic properties. For the carcinogenic effect, a genotoxic mechanism was considered possible, but was discounted by the European Food Safety Authority in 2004 based on new evidence. This allowed the derivation of
Sukhendu Maiti et al.
Journal of the American Chemical Society, 135(11), 4567-4572 (2013-03-07)
We present here, the design, synthesis, spectroscopic characterization, and in vitro biological assessment of a gemcitabine-coumarin-biotin conjugate (5). Probe 5 is a multifunctional molecule composed of a thiol-specific cleavable disulfide bond, a coumarin moiety as a fluorescent reporter, gemcitabine (GMC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service