Skip to Content
Merck
All Photos(1)

Key Documents

71383

Sigma-Aldrich

Sodium chloride

tested according to Ph. Eur.

Synonym(s):

Natrii chloridum, Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.7% (dried substance)
form:
solid
solubility:
water: soluble (358 g/l at 20 °C (68 °F))

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

vapor pressure

1.33 (1.00 mmHg) at 865 °C hPa

Assay

99.7% (dried substance)

form

solid

pH

7

mp

801 °C (lit.)

solubility

water: soluble (358 g/l at 20 °C (68 °F))

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurent Dortet et al.
Journal of clinical microbiology, 52(7), 2359-2364 (2014-04-25)
Multidrug-resistant Acinetobacter baumannii isolates, particularly those that produce carbapenemases, are increasingly reported worldwide. The biochemically based Carba NP test, extensively validated for the detection of carbapenemase producers among Enterobacteriaceae and Pseudomonas spp., has been modified to detect carbapenemase production in
Francis Kim et al.
JAMA, 311(1), 45-52 (2013-11-19)
Hospital cooling improves outcome after cardiac arrest, but prehospital cooling immediately after return of spontaneous circulation may result in better outcomes. To determine whether prehospital cooling improves outcomes after resuscitation from cardiac arrest in patients with ventricular fibrillation (VF) and
Alexander W Wyatt et al.
Genome biology, 15(8), 426-426 (2014-08-27)
Genomic analyses of hundreds of prostate tumors have defined a diverse landscape of mutations and genome rearrangements, but the transcriptomic effect of this complexity is less well understood, particularly at the individual tumor level. We selected a cohort of 25
Frédéric Lebrun-Julien et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(25), 8432-8448 (2014-06-21)
The mammalian target of rapamycin (mTOR) pathway integrates multiple signals and regulates crucial cell functions via the molecular complexes mTORC1 and mTORC2. These complexes are functionally dependent on their raptor (mTORC1) or rictor (mTORC2) subunits. mTOR has been associated with
Koji Tanaka et al.
Nature communications, 6, 6337-6337 (2015-02-27)
Pore-forming toxins (PFT) are water-soluble proteins that possess the remarkable ability to self-assemble on the membrane of target cells, where they form pores causing cell damage. Here, we elucidate the mechanism of action of the haemolytic protein fragaceatoxin C (FraC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service