Skip to Content
Merck
All Photos(1)

Key Documents

71230

Sigma-Aldrich

Thimerosal

97.0-101.0% (on dried substance, T)

Synonym(s):

2-(Ethylmercuriomercapto)benzoic acid sodium salt, Ethylmercurithiosalicylic acid sodium salt, Mercury-([o-carboxyphenyl]thio)ethyl sodium salt, Sodium ethylmercurithiosalicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(C2H5HgS)C6H4CO2Na
CAS Number:
Molecular Weight:
404.81
Beilstein:
8169555
EC Number:
MDL number:
UNSPSC Code:
51473202
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

97.0-101.0% (on dried substance, T)

form

powder

mp

234-237 °C (dec.) (lit.)

Mode of action

enzyme | inhibits

SMILES string

[Na+].CC[Hg]Sc1ccccc1C([O-])=O

InChI

1S/C7H6O2S.C2H5.Hg.Na/c8-7(9)5-3-1-2-4-6(5)10;1-2;;/h1-4,10H,(H,8,9);1H2,2H3;;/q;;2*+1/p-2

InChI key

RTKIYNMVFMVABJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Thimerosal is a sulfhydryl (thiol) oxidizing reagent capable of inhibiting a wide range of sulfhydryl-dependent enzymes and proteins such as PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na++K+ dependent ATPases.

Analysis Note

Solubility (EP/BP) Freely soluble in water, sparingly soluble or soluble in ethanol (96%), practically insoluble in ether and methylene cholride.

Other Notes

As part of the corporation global plan to support the Minamata Convention eliminating mercury-containing compounds, as of January 2021 customers will be required to provide an End-Use Declaration. Sales of thimerosal will be restricted for approved applications only.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ildiko R Dunay et al.
Antimicrobial agents and chemotherapy, 48(12), 4848-4854 (2004-11-25)
Acute therapy with pyrimethamine plus sulfadiazine is the treatment of choice for reactivated toxoplasmic encephalitis (TE). Acute therapy is followed by lifelong maintenance therapy (secondary prophylaxis) with the same drugs at lower dosages. The use of pyrimethamine plus sulfadiazine is
John M Feller et al.
The Journal of clinical endocrinology and metabolism, 93(9), 3560-3567 (2008-06-12)
Hyperinsulinism with islet cell hyperplasia is a frequent complication, of unknown cause, in hemolytic disease of the newborn, occurring in Rh(D)-positive infants of Rh-isoimmunized Rh(D)-negative mothers, but not in infants with other hemolytic disorders. We investigated the possibility that trans-placentally
Pan-Chyr Yang et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 111(6), 333-339 (2012-07-04)
This study evaluated the immune response elicited by two formulations of an AS03(A)-adjuvanted H5N1 A/Indonesia/05/2005 prepandemic influenza vaccine, developed using manufacturing processes with or without thiomersal. In addition, it also evaluated compliance to the Centre for Biologics Evaluation and Research
Dorota Mrozek-Budzyn et al.
Neurotoxicology and teratology, 34(6), 592-597 (2012-10-17)
Despite the common use of Thimerosal as a preservative in childhood vaccines since the 1930s, there are not many studies on ethylmercury toxicokinetics and toxicodynamics in infants. The knowledge of ethylmercury's potential adverse effects is derived mostly from parallel methylmercury
Alberto Eugenio Tozzi et al.
Developmental medicine and child neurology, 54(9), 843-848 (2012-05-18)
The aim of this article was to explore the effect of duration of breastfeeding on neurocognitive development. The long-term effect of breastfeeding on neurodevelopment was examined through a battery of neuropsychological tests in 1403 children (693 females, 710 males; mean

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service