Skip to Content
Merck

71022BC

Assurance® GDS Salmonella HET Tq Kit

suitable for PCR testing in Food and Beverages applications

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106302
NACRES:
NA.85
Suitability:
Salmonella enteritidis
Manufacturer/tradename:
Assurance GDS®, BioControl Systems 71022-100, BioControl Systems 71022-576ATM
Application(s):
food and beverages
pathogen testing
Technique(s):
PCR: suitable
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

GDS Salmonella HET Tq, BioControl, Molecular based PCR test for the detection of Salmonellla spp., Salmonella Heidelberg, Salmonella Enteritidis, and Salmonella typhimurium in raw poultry and environmental samples

Quality Segment

shelf life

6‑12 mo.

packaging

pkg of 100 tests

manufacturer/tradename

Assurance GDS®, BioControl Systems 71022-100, BioControl Systems 71022-576ATM

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

2-8°C

suitability

Salmonella enteritidis

General description

Assurance® GDS, genetic detection system, for Salmonella HET Tq is an automated nucleic acid amplification system for the detection of Salmonella Heidelberg, Salmonella Enteritidis and/or Salmonella Typhimurium (HET) in environmental boot swabs, poultry rinses, raw poultry, and raw ground poultry. The proprietary PickPen® IMS-based sample preparation procedure quickly and effectively captures Salmonella HET, separating the pathogen from inhibitory background materials ensuring valid PCR reactions for all sample types, including challenging food and environmental samples. Assurance® GDS PCR technology provides two additional levels of specificity through the use of both highly specific primers and probes to ensure accurate detection of Salmonella HET.

Application

Assurance® GDS Salmonella HET Tq is suitable for the detection of Salmonella spp. in environmental boot swabs, poultry rinses, raw poultry, and raw ground poultry.
PCR, Immunomagnetic Separation (IMS)

Other Notes

Each Assurance GDS for Salmonella HET Tq 100 kit contains the following:
  • Salmonella HET Amplification Tubes Tq (0.2 mL)
  • Salmonella HET Concentration Reagent
  • Resuspension Buffer Tq
  • Wash Solution

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10 - Combustible liquids

Signal Word

Danger

Hazard Codes

Hazard Classifications

ED ENV 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library