Skip to Content
Merck

69734

Multi Anion Standard 1 for IC

TraceCERT®, in water

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

composition

bromide, 20 mg/L , chloride, 10 mg/L , fluoride, 3 mg/L , nitrate, 20 mg/L , phosphate, 30 mg/L , sulfate, 20 mg/L

concentration

in water

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

multi-component solution

Application

Multi anion standard 1 for IC has been used as a multi-element analytical standard to determine the composition of ions in tomatoes using the ion chromatography (IC) technique.

Analysis Note

For the preparation of this CRM only materials of highest purity were used.

Other Notes

Visit the IC Portal to learn more

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Martina Paponov et al.
Frontiers in plant science, 10, 1656-1656 (2020-01-31)
We investigated the effect of supplemental LED inter-lighting (80% red, 20% blue; 70 W m-2; light period 04:00-22:00) on the productivity and physiological traits of tomato plants (Flavance F1) grown in an industrial greenhouse with high pressure sodium (HPS) lamps
Supplemental LED inter-lighting increases tomato fruit growth through enhanced photosynthetic light use efficiency and modulated root activity
Paponov M, et al.
Frontiers in Plant Science, 10, 1656-1656 (2019)



Global Trade Item Number

SKUGTIN
69734-100ML04061832786513