Skip to Content
Merck
All Photos(2)

Documents

69545

Supelco

Kaempferide

analytical standard

Synonym(s):

3,5,7-Trihydroxy-4′-methoxyflavone, KF, 4′-Methoxy-3,5,7-trihydroxyflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
Beilstein:
305378
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C2=C(O)C(=O)c3c(O)cc(O)cc3O2

InChI

1S/C16H12O6/c1-21-10-4-2-8(3-5-10)16-15(20)14(19)13-11(18)6-9(17)7-12(13)22-16/h2-7,17-18,20H,1H3

InChI key

SQFSKOYWJBQGKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kaempferide is a naturally occurring, O-methylated flavonol found in Kaempferia galanga. It exhibits a variety of pharmacological effects such as anti-inflammatory, anti-viral, anti-oxidant, anti-bacterial and antitumor properties.

Application

Kaempferide may be used as an analytical reference standard for the quantification of the analyte in propolis and traditional Chinese medicine Zuo Gui Wan using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: arnica silybum

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaempferide prevents titanium particle induced osteolysis by suppressing JNK activation during osteoclast formation
Jiao Z, et al.
Scientific Reports, 7(1) (2017)
Madoka Takashima et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 132, 110669-110669 (2019-07-13)
Propolis is a sticky dark-colored substance produced by honey bees and comprises resin, balsam, wax, essential and aromatic oils, pollen, and several other substances; it is used in food and beverages to improve health and prevent diseases. We studied the
Ehab Ghazy et al.
European journal of medicinal chemistry, 200, 112338-112338 (2020-06-05)
Histone modifying proteins, specifically histone deacetylases (HDACs) and bromodomains, have emerged as novel promising targets for anticancer therapy. In the current work, based on available crystal structures and docking studies, we designed dual inhibitors of both HDAC6/8 and the bromodomain
Selective analysis of phenolic compounds in propolis by HPLC?MS/MS
Medana C, et al.
Phytochemical Analysis, 19(1), 32-39 (2008)
A combined HPLC-PDA and HPLC-MS method for quantitative and qualitative analysis of 10 major constituents in the traditional Chinese medicine Zuo Gui Wan
Liang X, et al.
Journal of Pharmaceutical and Biomedical Analysis, 49(4), 931-936 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service