Skip to Content
Merck
All Photos(1)

Key Documents

68228

Supelco

2-Decanone

analytical standard

Synonym(s):

Methyl octyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7COCH3
CAS Number:
Molecular Weight:
156.27
Beilstein:
1747463
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

>1 (vs air)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.425 (lit.)
n20/D 1.425

bp

211 °C (lit.)

mp

3.5 °C (lit.)

density

0.825 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCC(C)=O

InChI

1S/C10H20O/c1-3-4-5-6-7-8-9-10(2)11/h3-9H2,1-2H3

InChI key

ZAJNGDIORYACQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Decanone can be obtained via liquid-phase Wacker oxidation of 1-decene in the presence of an efficient catalyst like Pd60 (Pd- palladium) nanocluster immobilized on titanium oxide support and cuprous chloride/H2O.[1]
2-Decanone is a constituent of the essential oil of Ruta chalepensis L., which is used in the production of food, drugs and perfumery.[2]

Application

2-Decanone may be used as an analytical standard for the determination of the analyte in the essential oil of Ruta chalepensis L.,[2] milk[3] and cheese[4] by chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct thermal desorption in the analysis of cheese volatiles by gas chromatography and gas chromatography-mass spectrometry: comparison with simultaneous distillation-extraction and dynamic headspace
Valero E, et al.
Journal of Chromatographic Science, 39(6), 222-228 (2001)
Nanoscale palladium cluster immobilized on a TiO2 Surface as an efficient catalyst for liquid-phase Wacker oxidation of higher terminal olefins
Choi M-K, et al.
Chemistry Letters (Jpn), 32(2), 180-181 (2003)
Chemical composition of the essential oil of Ruta chalepensis L: Influence of drying, hydro-distillation duration and plant parts
Mejri J, et al.
Industrial Crops and Products, 32(3), 671-673 (2010)
Quantitative determination of thermally derived off-flavor compounds in milk using solid-phase microextraction and gas chromatography
Vazquez-Landaverde PA, et al.
Journal of Dairy Science, 88(11), 3764-3772 (2005)
Peter S Toth et al.
The Analyst, 140(6), 1947-1954 (2015-02-11)
Two-phase voltammetry has been carried out using a reverse cell configuration, i.e. with the lower density organic solvent on the top of the aqueous solution in the cell, where the organic solvents contain either nitrile or ketone functional groups. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service