Skip to Content
Merck

Skip To

67372

Melting point standard 283-286°C

analytical standard

Synonym(s):

Anthraquinone, ATQ

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H8O2
CAS Number:
Molecular Weight:
208.21
UNSPSC Code:
20121904
NACRES:
NA.24
PubChem Substance ID:
EC Number:
201-549-0
Beilstein/REAXYS Number:
390030
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

7.16 (vs air)

vapor pressure

1 mmHg ( 190 °C)

shelf life

limited shelf life, expiry date on the label

bp

379-381 °C (lit.)

mp

283-286 °C (±0.3°C), 284-286 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

O=C1c2ccccc2C(=O)c3ccccc13

InChI

1S/C14H8O2/c15-13-9-5-1-2-6-10(9)14(16)12-8-4-3-7-11(12)13/h1-8H

InChI key

RZVHIXYEVGDQDX-UHFFFAOYSA-N

General description

Melting point standard 283-286 °C, anthraquinone is an analytical standard, suitable for the day-to-day calibration of melting point apparatus to guarantee its accuracy, in accordance with local, national and international standards laboratories.
The mp value is recorded as an average of 6 to 12 measurements with a Büchi B-545 equipment that is calibrated against primary standards. Melting point is determined by Capillary method, as described in Ph. Eur. 2.2.14.

Application

Anthraquinone melting point standard 283-286 °C is an mp standard used for physicochemical characterization.
This product also features as a reference standard in thermal property investigations.

Features and Benefits

  • Melting point calibration standard traceable to primary standards (LGC, London)
  • Grade: Analytical Standard
  • Melting point validated in the thermodynamic mode of analysis
  • Standard deviation up to ± 0.3 °C
  • Available with certificates of analysis and safety data sheet


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
421837617050296
format

neat

format

neat

format

neat

format

neat

bp

379-381 °C (lit.)

bp

-

bp

249 °C (lit.)

bp

305 °C (lit.)

mp

283-286 °C (±0.3°C), 284-286 °C (lit.)

mp

182-184 °C (±0.3°C), 182-185 °C (lit.)

mp

121-123 °C (±0.3°C), 121-125 °C (lit.)

mp

47-49 °C (±0.3°C), 47-51 °C (lit.)

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

application(s)

food and beverages
pharmaceutical

application(s)

food and beverages
pharmaceutical

application(s)

food and beverages
pharmaceutical

application(s)

food and beverages
pharmaceutical

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label


pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

482.0 °F - closed cup

flash_point_c

250 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters