Skip to Content
Merck
All Photos(1)

Documents

65943

Supelco

N-Methyl-bis(trifluoroacetamide)

for GC derivatization, LiChropur, ≥99.0% (GC)

Synonym(s):

MBTFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CON(CH3)COCF3
CAS Number:
Molecular Weight:
223.07
Beilstein:
1878402
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥99.0% (GC)

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.346 (lit.)
n20/D 1.346

bp

121-122 °C (lit.)

density

1.547 g/mL at 25 °C

SMILES string

CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F

InChI

1S/C5H3F6NO2/c1-12(2(13)4(6,7)8)3(14)5(9,10)11/h1H3

InChI key

AWGBWLXGUPTXHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methyl-bis(trifluoroacetamide) is a common reagent used in the alkylation process, which targets highly polar, multi-functional compounds such as carbohydrates and amino acids.

Application

N-Methyl-bis(trifluoroacetamide) may be used as an on-column derivatization agent for the determination of amphetamine-type stimulants in biological samples, and ketamine, norketamine, dehydronorketamine in urine samples using gas chromatography coupled to mass spectrometry technique.
It is a derivatization agent in GC-MS technique used as an alternative to trifluoroacetyl anhydride (TFAA) for trifluoroacylating primary and secondary amines, and the hydroxyl group under mild non-acidic conditions.
Reagent for introducing trifluoroacetyl group. Suitable for the derivatization of alcohols, amines, amino acids, carbohydrates(mono-,di-,tri- and tetrasaccharides, carboxylic acids, hydroxyamines, 3-hydroxy-1-nitrosopyrrolidine, indolalkylamine, phenolalkylamines, pyridoxal (and other B6 vitamers), and thiols.
Reagent for introducing trifluoroacetyl group.

Other Notes

Reagent for trifluoroacetyl, N-trimethylsilyl-N-tifluoroacetyl and trimethylsilyl.
Reagent for trifluoroacetylations under mild conditions

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a two-step injector for GC?MS with on-column derivatization, and its application to the determination of amphetamine-type stimulants (ATS) in biological specimens
Miki A, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 865(1-2), 25-32 (2008)
K. Blau et al.
Handbook of Derivatives for Chromatography, 131-131 (1977)
Quantitative detection of ketamine, norketamine, and dehydronorketamine in urine using chemical derivatization followed by gas chromatography?mass spectrometry
Huang K-M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 820(2), 165-173 (2005)
[Acylation with bis(acylamides). N-methyl-bis(trifluoroacetamide) and bis(trifluoroacetamide), two new reagents for trifluoroacetylation].
M Donike
Journal of chromatography, 78(2), 273-279 (1973-04-25)
Derivatization reactions and reagents for gas chromatography analysis
Advanced Gas Chromatography: Progress in Agricultural, Biomedical and Industrial Applications, 83-108 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service