Skip to Content

61397

Hybriscan D kit

Yeast, suitable for food and beverages, microbiology and in situ hybriziation

Synonym(s):

FastScan Yeast, Yeast HybriScanD

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85
Suitability:
molds, yeasts
Manufacturer/tradename:
(HybriScan)
Application(s):
food and beverages, microbiology
Technique(s):
microbe id | in situ hybridization: suitable
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

HybriScanD Yeast, suitable for microbiology

manufacturer/tradename

(HybriScan)

Quality Segment

technique(s)

microbe id | in situ hybridization: suitable

application(s)

food and beverages, microbiology

storage temp.

2-8°C

suitability

molds, yeasts

General description

HybriScan D kits detect and simultaneously identify microorganisms using rRNA as the detection target. The principle is based on in situ sandwich hybridization with two specific probes. As the rRNA of dead cells is degraded in the presence of RNase, only living cells are detected. Each cell has several thousand rRNA copies, so neither PCR nor a polymerase is needed for this technology, which means that this molecular biology test is not inhibited by the sample matrix, for example food, beverages and water. We recommend a 24-hour enrichment in Tomato Juice Broth (Cat. No. 17218)

Features and Benefits

  • Sensitivity (after pre enrichment): 1-10 cfu/L
  • Sensitivity (cfu/assay): 500

Legal Information

HybriScan is a trademark of ScanBec GmbH


Still not finding the right product?

Explore all of our products under HybriScanD Yeast


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library