Skip to Content
Merck
All Photos(1)

Key Documents

523321

Supelco

Poly(2-vinylpyridine)

analytical standard, average Mw 37,500 (Typical), average Mn 35,000 (Typical)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

mol wt

average Mn 35,000 (Typical)
average Mw 37,500 (Typical)

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

InChI

1S/C7H7N/c1-2-7-5-3-4-6-8-7/h2-6H,1H2

InChI key

KGIGUEBEKRSTEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Synthesis and Structural Insight into Poly(dimethylsiloxane)-b-Poly(2-vinylpyridine) Copolymers: This study delves into the synthesis of poly(dimethylsiloxane)-b-poly(2-vinylpyridine) copolymers, highlighting their structural characteristics and potential applications in creating hybrid materials with unique properties (Avgeropoulos et al., 2023).


  • pH Tunable Patterning of Quantum Dots: Research discusses the utilization of poly(2-vinylpyridine) in pH-sensitive patterning for quantum dot arrays, offering a versatile approach for developing advanced photonic devices (Onses et al., 2024).


  • Dynamic Photonic Janus Colloids with Axially Stacked Structural Layers: The study presents the application of poly(2-vinylpyridine) in the formation of dynamic photonic Janus colloids, which could lead to innovative solutions in sensor technologies and display systems (Ku et al., 2024).


  • Polyhedral Colloidal Clusters Assembled from Amphiphilic Nanoparticles in Deformable Droplets: Demonstrates the role of poly(2-vinylpyridine) in assembling complex colloidal structures, which may be crucial for the development of novel biomaterials and drug delivery systems (Jiang et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianying Jiang et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 24(37), 375109-375109 (2012-08-24)
Here we investigate the flow properties of suspensions of dicolloidal particles composed of interpenetrating spheres where one sphere is rich in polystyrene and the second is rich in poly 2-vinyl pyridine. The synthesis method is designed to create both anisotropic
Keewook Paeng et al.
The Journal of chemical physics, 134(2), 024901-024901 (2011-01-19)
A temperature-ramping anisotropy measurement is introduced as an efficient way to study molecular motion in polymer glasses. For these experiments, fluorescent molecules were dispersed in the polymer glass and the reorientation of these dyes was used as a probe of
Natalia Kocharova et al.
Langmuir : the ACS journal of surfaces and colloids, 23(6), 3363-3371 (2007-02-13)
We report the efficient aqueous dispersion of pristine HiPco single-walled carbon nanotubes (SWNTs) with ionic liquid (IL)-based surfactants 1-dodecyl-3-methylimidazolium bromide (1) and 1-(12-mercaptododecyl)-3-methylimidazolium bromide (2), the thiolation of nanotube sidewalls with 2, and the controlled self-assembly of positively charged SWNT-1,2
Weihua Tao et al.
Journal of hazardous materials, 175(1-3), 111-116 (2009-11-03)
In the present study, the performance, behavior and mechanism of synthetic poly(2-vinylpyridine) resin (WH-225) adsorbing 5-sodiosulfoisophthalic acids (SIPA) from the aqueous solution were investigated, and two commercial adsorbents, namely, hypercrosslinked adsorbent NDA-100 and macroporous adsorbent XAD-4 were employed as reference.
Mohammad Changez et al.
Advanced materials (Deerfield Beach, Fla.), 24(24), 3253-3257 (2012-05-19)
The reaction between atactic poly(2-vinylpyridine) and 1,4-dibromobutane leads to formation of long-range 3D molecular ordering in polymer chains mainly because the side group (pyridine) of the polymer chain changes to a syndotactic configuration. This may enable the production of functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service