Skip to Content
Merck
All Photos(1)

Key Documents

51681

Supelco

Silychristin

analytical standard

Synonym(s):

Silymarin II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(ccc1O)[C@@H]2Oc3c(O)cc(cc3[C@H]2CO)[C@H]4Oc5cc(O)cc(O)c5C(=O)[C@@H]4O

InChI

1S/C25H22O10/c1-33-18-6-10(2-3-15(18)28)23-14(9-26)13-4-11(5-17(30)25(13)35-23)24-22(32)21(31)20-16(29)7-12(27)8-19(20)34-24/h2-8,14,22-24,26-30,32H,9H2,1H3/t14-,22+,23+,24-/m1/s1

InChI key

BMLIIPOXVWESJG-LMBCONBSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silychristin is one of the congeners of flavonolignan silybin. It is usually derived from silymarin, which is extracted from the fruits of Silybum marianum (L.) Gaertn, and Asteraceae (Carduus marianus L.; milk thistle).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Silychristin may be used as an analytical reference standard for the determination of silychristin in:
  • Herbal extracts, fruit extracts, and seed extracts of Silybum marianum by ultra-high performance liquid chromatography with diode array detection (UHPLC-DAD) and methanol extraction coupled to HPLC equipped with ultraviolet array detection (UVAD).
  • Milk thistle seeds by HPLC-UV.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacokinetics of pure silybin diastereoisomers and identification of their metabolites in rat plasma.
Marhol P, et al.
Journal of functional foods, 14, 570-580 (2015)
Optimization and single-laboratory validation of a method for the determination of flavonolignans in milk thistle seeds by high-performance liquid chromatography with ultraviolet detection.
Mudge E, et al.
Analytical and Bioanalytical Chemistry, 407(25), 7657-7666 (2015)
Head-to-head comparison of ultra-high-performance liquid chromatography with diode array detection versus quantitative nuclear magnetic resonance for the quantitative analysis of the Silymarin complex in Silybum marianum fruit extracts.
Cheilari A, et al.
Journal of Agricultural and Food Chemistry, 64(7), 1618-1626 (2016)
Lijun Duan et al.
Applied biochemistry and biotechnology, 113-116, 559-568 (2004-04-01)
Hot water is attracting attention as an extraction solvent in the recovery of compounds from plant material as the search for milder and "greener" solvents intensifies. The use of hot water as an extraction solvent for milk thistle at temperatures
Ludovico Abenavoli et al.
Phytotherapy research : PTR, 24(10), 1423-1432 (2010-06-22)
Silybum marianum or milk thistle (MT) is the most well-researched plant in the treatment of liver disease. The active complex of MT is a lipophilic extract from the seeds of the plant and is composed of three isomer flavonolignans (silybin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service