Skip to Content
Merck
All Photos(1)

Documents

51490

Millipore

Hektoen Enteric Agar

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

acid fuchsin, 0.1 g/L
agar, 15.0 g/L
ammonium ferric citrate, 1.5 g/L
bile salts, 9 g/L
bromothymol blue, 0.065 g/L
lactose, 12 g/L
mixed peptone, 12 g/L
salicin, 2 g/L
sodium chloride, 5 g/L
sodium thiosulfate, 5 g/L
sucrose, 12 g/L
yeast extract, 3 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.5±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages
water monitoring

microbiology

suitability

selective and differential for Salmonella spp.
selective and differential for Shigella spp.
selective and differential for bacteria (General Media)

Application

Hektoen enteric agar is a type of primary standard media, which may be commonly used in the selective detection and isolation of Salmonella in food microbiology labs and in human stool specimens.

Preparation Note

Suspend 76.67 g in 1 litre distilled water and let it soak. Heat up by constant stirring until boiling. Cool to 55 - 60 °C and pour into sterile plates. Do not autoclave. The Medium is very thermolabile and thus overheating should be avoided.

Other Notes

Plating medium for the isolation of enteric pathogens; Isolation of Shigellae

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Entis P
Food Microbiology: The Laboratory (2002)
Comparison of four chromogenic media and Hektoen agar for detection and presumptive identification of Salmonella strains in human stools
Perez.MJ, et al.
Journal of Clinical Microbiology, 41, 1130-1134 (2003)
S. King et al.
Applied Microbiology, 16, 557-557 (1968)
W I Taylor et al.
Applied microbiology, 21(1), 32-37 (1971-01-01)
Two enrichment broths and four plating media were compared for efficiency of detection of enteric pathogens from 1,597 stool specimens. Of 170 salmonellae isolated from the composite of all methods, direct streaking yielded but 54%, whereas enrichment in gram-negative broth
S King et al.
Applied microbiology, 16(4), 579-581 (1968-04-01)
During this study, 2,855 stool specimens from patients at Cook County Hospital were cultured for enteric pathogens. Hektoen Enteric Agar (HE) was compared with E M B and S S Agars by replicate samplings with both direct and indirect methods.

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service