Skip to Content
Merck

Skip To

50866

[6]-Gingerol

analytical standard

Synonym(s):

3-Decanone, 5-hydroxy-1-(4-hydroxy-3-methoxyphenyl)-, (5S)-, 5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-decanone, 6-Gingerol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H26O4
CAS Number:
Molecular Weight:
294.39
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥94.0% (HPLC)

shelf life

expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCC[C@H](O)CC(=O)CCc1ccc(O)c(OC)c1

InChI

1S/C17H26O4/c1-3-4-5-6-14(18)12-15(19)9-7-13-8-10-16(20)17(11-13)21-2/h8,10-11,14,18,20H,3-7,9,12H2,1-2H3/t14-/m0/s1

InChI key

NLDDIKRKFXEWBK-AWEZNQCLSA-N

General description

[6]-Gingerol is a naturally occurring plant phenoland an active pungent constituent found in the rhizome of ginger, which is known to possess anti-inflammatory, anti-tumor and antioxidant properties and can hence, serve as a potential candidate in the treatment of cancer.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Bioactive compound found in ginger (Zingiber officinale) with antioxidant activity, which functions as an anti-inflammatory and antitumor agent. [6]-Gingerol down regulates proinflammatory cytokine release by macrophages. It has been shown to inhibit COX-2 expression by blocking the activation of p38 MAP kinase and NF-κB in phorbol ester-stimulated mouse skin.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: zingiber

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHL89201G1046PHL89202
application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

food and beverages

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

application(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

Quality Level

100

Quality Level

-

Quality Level

200

Quality Level

-

grade

analytical standard

grade

primary reference standard

grade

-

grade

primary reference standard

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

assay

≥94.0% (HPLC)

assay

≥90.0% (HPLC)

assay

≥98% (HPLC)

assay

≥95.0% (HPLC)


Still not finding the right product?

Explore all of our products under [6]-Gingerol


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters