Skip to Content
Merck
All Photos(1)

Documents

50475

Supelco

Benzyl acetate

analytical standard

Synonym(s):

Acetic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2C6H5
CAS Number:
Molecular Weight:
150.17
Beilstein:
1908121
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

23 mmHg ( 110 °C)

Assay

≥99.7% (GC)

autoignition temp.

862 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.502 (lit.)
n20/D 1.503

bp

206 °C (lit.)

mp

−51 °C (lit.)

density

1.054 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)OCc1ccccc1

InChI

1S/C9H10O2/c1-8(10)11-7-9-5-3-2-4-6-9/h2-6H,7H2,1H3

InChI key

QUKGYYKBILRGFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl acetates are generally used as flavoring agents and preservatives. They find applications in foods and beverages, drugs, cosmetics, pharmaceuticals, etc.

Application

Benzyl acetate has been used as a reference standard for the determination of the analyte in e-cigarette refill solutions as flavoring additives and in muskmelon and watermelon fruits as aromatic agents by gas chromatography-mass spectrometry (GC-MS), and in scented consumer products as odor compounds by gas chromatography-olfactometry (GC-O).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of odour compounds from scented consumer products using gas chromatography-mass spectrometry and gas chromatography-olfactometry
Bartsch J, et al.
Analytica Chimica Acta, 904(5), 98-106 (2016)
Quantification of prominent volatile compounds responsible for muskmelon and watermelon aroma by purge and trap extraction followed by gas chromatography-mass spectrometry determination
Fredes A, et al.
Food Chemistry, 190(5), 689-700 (2016)
Comprehensive determination of flavouring additives and nicotine in e-cigarette refill solutions. Part II: Gas-chromatography-mass spectrometry analysis
Aszyk J, et al.
Journal of Chromatography A, 1517(5), 156-164 (2017)
J Caldwell
Teratogenesis, carcinogenesis, and mutagenesis, 13(4), 185-190 (1993-01-01)
The mouse lymphoma assay is commonly included in batteries of tests for the mutagenic and carcinogenic potential of chemicals. Unlike other bacterial and mammalian cell tests, it has an unacceptably high incidence of false positive results. This paper reviews data
C Debonneville et al.
Analytical chemistry, 74(10), 2345-2351 (2002-06-01)
A multisniffing system has been developed to allow three panelists to simultaneously participate in a GC-olfactometric analysis. This device, associated with a computerized data treatment, allows shortening CHARM and GC-"SNIF' analyses to less than 1 week and less than 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service