Skip to Content
Merck
All Photos(1)

Key Documents

46754

Supelco

Streptomycin sesquisulfate hydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H39N7O12 · 1.5H2O4S · xH2O
CAS Number:
Molecular Weight:
728.69 (anhydrous basis)
EC Number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
clinical
environmental
forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

O.OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.CN[C@H]1[C@H](O)[C@@H](O)[C@H](CO)O[C@H]1O[C@H]2[C@@H](O[C@@H](C)[C@]2(O)C=O)O[C@@H]3[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]3NC(N)=N.CN[C@H]4[C@H](O)[C@@H](O)[C@H](CO)O[C@H]4O[C@H]5[C@@H](O[C@@H](C)[C@]5(O)C=O)O[C@@H]6[C@@H](O)[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@H]6NC(N)=N

InChI

1S/2C21H39N7O12.3H2O4S.H2O/c2*1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35;3*1-5(2,3)4;/h2*4-18,26,29,31-36H,3H2,1-2H3,(H4,22,23,27)(H4,24,25,28);3*(H2,1,2,3,4);1H2/t2*5-,6-,7-,8+,9

InChI key

PJKBJPAUGKHYQJ-QXKUBQGSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noémi László et al.
Food chemistry, 267, 178-186 (2018-06-24)
The aim of our study was to examine the degradation of veterinary antibiotics in milk during boiling. Raw cow milk samples were fortified with the target compounds and boiled for various short time-intervals prevailing in household practice. Antibiotic concentrations were
Aleksandar Siljanoski et al.
The Journal of dairy research, 85(3), 321-326 (2018-07-04)
The purpose of this study was to determine tetracycline and other antimicrobial residues in milk from dairy cows treated for clinical mastitis. Data on cow health and drugs used were obtained from the farm veterinarians. A milk sample from each
P Bharali et al.
Colloids and surfaces. B, Biointerfaces, 103, 502-509 (2012-12-25)
In the present study, the interaction of rhamnolipid produced by Pseudomonas aeruginosa OBP1 with the cell surfaces of Staphylococcus aureus (MTCC 3160) and Klebsiella pneumoniae (MTCC 618) were studied. Rhamnolipid concentration below critical micelle concentration (CMC) did not exhibit significant
Prashant K Khade et al.
Journal of molecular biology, 425(20), 3778-3789 (2013-04-02)
Accurate tRNA selection by the ribosome is essential for the synthesis of functional proteins. Previous structural studies indicated that the ribosome distinguishes between cognate and near-cognate tRNAs by monitoring the geometry of the codon-anticodon helix in the decoding center using
Fred Stephen Sarfo et al.
PLoS neglected tropical diseases, 7(3), e2101-e2101 (2013-03-22)
Mycobacterium ulcerans infection causes a neglected tropical disease known as Buruli ulcer that is now found in poor rural areas of West Africa in numbers that sometimes exceed those reported for another significant mycobacterial disease, leprosy, caused by M. leprae.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service